Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080378.1 | NP_001073847.1 | 6 | 979 | |
NM_001080379.1 | NP_001073848.1 | 5 | 1021 | ||
NM_152410.2 | NP_689623.2 | 7 | 1096 | ||
GenBank mRNA | AF546872.1 | - | 5 | 1021 | |
AK057286.1 | - | 7 | 1096 | ||
BC030642.2 | - | 5 | 991 | 4823539 | |
BC044227.1 | - | 6 | 966 | 5187302 |