Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173854.5 | NP_776253.3 | 8 | 2076 | |
XM_005245069.2 | XP_005245126.1 | 8 | 1752 | ||
GenBank mRNA | AJ514402.1 | - | 7 | 1163 | |
AK125075.1 | - | 7 | 837 | ||
AK128069.1 | - | 7 | 1329 | ||
AK314732.1 | - | 7 | 1234 | ||
AL831974.1 | - | 8 | 1912 | ||
AL832001.1 | - | 8 | 1912 | ||
AL832362.1 | - | 8 | 1912 | ||
BC136303.1 | - | 7 | 1557 | 9020290 | |
BX641054.1 | - | 8 | 1914 | ||
BX648979.1 | - | 8 | 1950 |