Gene Transcripts
- Gene Symbol
- LINC01142
- Entrez Gene ID
- 284688
- Gene Name
- long intergenic non-protein coding RNA 1142
- Gene Aliases
- Chromosome Location
- Chr. 1: 170271405 - 170284208 on Build GRCh38
- UniGene ID
- Hs.135668
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026957.1 |
|
3 |
716 |
|
GenBank mRNA |
BC033381.2 |
- |
2 |
646 |
|
BC125144.1 |
- |
1 |
113 |
|
View Full Product Details