Gene Transcripts

Gene Symbol
LOC339059
Entrez Gene ID
339059
Gene Name
uncharacterized LOC339059
Gene Aliases
Chromosome Location
Chr. 16: 88742767 - 88745748 on Build GRCh38
UniGene ID
Hs.449817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120387.1 1 571
GenBank mRNA AK097767.1 - 1 571
BC111769.1 - 1 517 40025355

View Full Product Details