Gene Transcripts
- Gene Symbol
- LOC100419583
- Entrez Gene ID
- 100419583
- Gene Name
- ring finger protein 4 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 15: 44728988 - 44732851 on Build GRCh38
- UniGene ID
- Hs.489254
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125376.1 |
|
1 |
26 |
|
GenBank mRNA |
DC350419.1 |
- |
1 |
26 |
|
View Full Product Details