Gene Transcripts

Gene Symbol
PRAC2
Entrez Gene ID
360205
Gene Name
prostate cancer susceptibility candidate 2
Gene Aliases
C17orf93, HOXB-AS5, HOXB13-AS1, NCRNA00253
Chromosome Location
Chr. 17: 48720802 - 48724758 on Build GRCh38
UniGene ID
Hs.236557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282276.1 NP_001269205.1 1, 2 88
XM_011524747.2 XP_011523049.1 1, 2 80
GenBank mRNA AI762586.1 - 1, 2 419
BX106051.1 - 1, 2 88

View Full Product Details