Gene Transcripts

Gene Symbol
MYO15B
Entrez Gene ID
80022
Gene Name
myosin XVB
Gene Aliases
MYO15BP
Chromosome Location
Chr. 17: 75587545 - 75626850 on Build GRCh38
UniGene ID
Hs.390817
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001309242.1 NP_001296171.1
56 8268
XM_017025120.1 XP_016880609.1
58 7975
XM_017025121.1 XP_016880610.1
58 7960
XM_017025122.1 XP_016880611.1
58 7939
XM_017025123.1 XP_016880612.1
58 7911
XM_017025124.1 XP_016880613.1
57 7849
XM_017025125.1 XP_016880614.1
56 7698
XM_017025126.1 XP_016880615.1
53 7440
XM_017025127.1 XP_016880616.1
56 7283
XM_017025128.1 XP_016880617.1
57 7029
XM_017025129.1 XP_016880618.1
56 6261
XM_017025130.1 XP_016880619.1
55 6217
XM_017025131.1 XP_016880620.1
54 6656
XM_017025132.1 XP_016880621.1
53 6597
XM_017025133.1 XP_016880622.1
51 6122
XM_017025134.1 XP_016880623.1
52 6483
XM_017025135.1 XP_016880624.1
46 5139
XM_017025136.1 XP_016880625.1
46 5488
XM_017025137.1 XP_016880626.1
43 5033
XM_017025138.1 XP_016880627.1
38 4529
XM_017025139.1 XP_016880628.1
39 4330
XM_017025140.1 XP_016880629.1
36 4143
XM_017025143.1 XP_016880632.1
31 3420
XM_017025144.1 XP_016880633.1
52 7903
GenBank mRNA AF418292.1 -
31 3878
AF418293.1 -
32 3484
AK026339.1 -
8 583
AK096869.1 -
8 722
AL713730.1 -
7 477
BC027875.1 - LIT3316
MYO15BP
8 972

View Full Product Details