Gene Transcripts
- Gene Symbol
- PELI3
- Entrez Gene ID
- 246330
- Gene Name
- pellino E3 ubiquitin protein ligase family member 3
- Gene Aliases
- Chromosome Location
- Chr. 11: 66466327 - 66477337 on Build GRCh38
- UniGene ID
- Hs.656875
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001098510.1 |
NP_001091980.1
|
5, 6 |
731 |
|
NM_001243135.1 |
NP_001230064.1
|
5, 6 |
570 |
|
NM_001243136.1 |
NP_001230065.1
|
4, 5 |
578 |
|
NM_145065.2 |
NP_659502.2
|
6, 7 |
803 |
|
XM_011544884.2 |
XP_011543186.1
|
6, 7 |
806 |
|
XM_017017465.1 |
XP_016872954.1
|
4, 5 |
555 |
|
GenBank mRNA |
AF487456.1 |
- |
5, 6 |
639 |
|
AF487457.1 |
- |
4, 5 |
567 |
|
AK074201.1 |
- |
6, 7 |
741 |
|
AK075253.1 |
- |
2, 3 |
243 |
|
AK094060.1 |
- |
5, 6 |
570 |
|
AL834395.1 |
- |
6, 7 |
750 |
|
BC021256.2 |
- |
6, 7 |
744 |
3535985 |
BC025723.1 |
- |
5, 6 |
652 |
5198999 |
BX341733.2 |
- |
4, 5 |
515 |
|
View Full Product Details