Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000173.6 | NP_000164.5 | 2 | 692 | |
GenBank mRNA | BC027955.1 | - | 2 | 670 | |
J02940.1 | - | 2 | 637 | ||
KC120774.1 | - | 2 | 670 | ||
KC120775.1 | - | 2 | 670 | ||
KC120776.1 | - | 2 | 670 |