Gene Transcripts

Gene Symbol
SMIM10
Entrez Gene ID
644538
Gene Name
small integral membrane protein 10
Gene Aliases
CXorf69
Chromosome Location
Chr. X: 134990938 - 134992473 on Build GRCh38
UniGene ID
Hs.42239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163438.1 NP_001156910.1 1 393
GenBank mRNA BC008642.1 - 1 393

View Full Product Details