Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102397278-102397824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF1
Entrez Gene ID
3479
Gene Name
insulin like growth factor 1
Gene Aliases
IGF-I, IGFI, MGF
UniGene
Hs.160562
Gene Chromosome Location
Chr.12: 102395867-102481839 on Build GRCh38
Transcripts
NM_000618.4 NM_001111283.2 NM_001111284.1 XM_017019262.1 XM_017019263.1

SNP Detail

SNP ID Chromosome Location
rs5742711 Chr.12: 102397305-102397305 on Build GRCh38
rs901224963 Chr.12: 102397685-102397685 on Build GRCh38
rs750216810 Chr.12: 102397717-102397717 on Build GRCh38
rs949292668 Chr.12: 102397323-102397323 on Build GRCh38
rs758592067 Chr.12: 102397538-102397538 on Build GRCh38
rs886048863 Chr.12: 102397320-102397320 on Build GRCh38
rs769629790 Chr.12: 102397592-102397592 on Build GRCh38
rs886717144 Chr.12: 102397603-102397603 on Build GRCh38
rs936485863 Chr.12: 102397442-102397442 on Build GRCh38
rs942455423 Chr.12: 102397537-102397537 on Build GRCh38
rs5742709 Chr.12: 102397749-102397749 on Build GRCh38
rs886048866 Chr.12: 102397567-102397567 on Build GRCh38
rs756085575 Chr.12: 102397764-102397764 on Build GRCh38
rs908188642 Chr.12: 102397431-102397431 on Build GRCh38
rs1054943063 Chr.12: 102397492-102397492 on Build GRCh38
rs796764972 Chr.12: 102397523-102397524 on Build GRCh38
rs993885014 Chr.12: 102397574-102397574 on Build GRCh38
rs374230240 Chr.12: 102397413-102397413 on Build GRCh38
rs952939127 Chr.12: 102397421-102397421 on Build GRCh38
rs70961706 Chr.12: 102397768-102397768 on Build GRCh38
rs576692277 Chr.12: 102397686-102397686 on Build GRCh38
rs953019192 Chr.12: 102397710-102397710 on Build GRCh38
rs1005782985 Chr.12: 102397726-102397726 on Build GRCh38
rs984791277 Chr.12: 102397428-102397428 on Build GRCh38
rs187947737 Chr.12: 102397343-102397343 on Build GRCh38
rs1006333433 Chr.12: 102397609-102397609 on Build GRCh38
rs961358519 Chr.12: 102397437-102397437 on Build GRCh38
rs946642309 Chr.12: 102397542-102397542 on Build GRCh38
rs556494579 Chr.12: 102397661-102397661 on Build GRCh38
rs927731868 Chr.12: 102397471-102397471 on Build GRCh38
rs886048865 Chr.12: 102397527-102397527 on Build GRCh38
rs560296866 Chr.12: 102397787-102397787 on Build GRCh38
rs752869775 Chr.12: 102397472-102397472 on Build GRCh38
rs1016400820 Chr.12: 102397667-102397668 on Build GRCh38
rs975517343 Chr.12: 102397354-102397354 on Build GRCh38
rs900845447 Chr.12: 102397562-102397562 on Build GRCh38
rs981880658 Chr.12: 102397453-102397453 on Build GRCh38
rs923834629 Chr.12: 102397746-102397746 on Build GRCh38
rs765324641 Chr.12: 102397407-102397407 on Build GRCh38
rs541590141 Chr.12: 102397770-102397770 on Build GRCh38
rs1028416673 Chr.12: 102397704-102397704 on Build GRCh38
rs542494292 Chr.12: 102397744-102397744 on Build GRCh38
rs1024940718 Chr.12: 102397689-102397689 on Build GRCh38
rs1026523001 Chr.12: 102397594-102397594 on Build GRCh38
rs886048864 Chr.12: 102397428-102397428 on Build GRCh38
rs1041033812 Chr.12: 102397678-102397678 on Build GRCh38
rs1042723142 Chr.12: 102397567-102397567 on Build GRCh38
rs5742710 Chr.12: 102397524-102397525 on Build GRCh38

View Full Product Details