Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102402188-102402729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF1
Entrez Gene ID
3479
Gene Name
insulin like growth factor 1
Gene Aliases
IGF-I, IGFI, MGF
UniGene
Hs.160562
Gene Chromosome Location
Chr.12: 102395867-102481839 on Build GRCh38
Transcripts
NM_000618.4 NM_001111283.2 NM_001111284.1 XM_017019262.1 XM_017019263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4548799 c.456G>C p.R152S Chr.12: 102402513-102402513 on Build GRCh38
3772228 c.444C>G p.N148K Chr.12: 102402525-102402525 on Build GRCh38
7191390 c.419A>G p.N140S Chr.12: 102402550-102402550 on Build GRCh38
4435350 c.403G>A p.E135K Chr.12: 102402566-102402566 on Build GRCh38
6071199 c.462G>T p.*154Y Chr.12: 102402507-102402507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201878788 Chr.12: 102402245-102402246 on Build GRCh38
rs1027850670 Chr.12: 102402250-102402250 on Build GRCh38
rs373139385 Chr.12: 102402383-102402383 on Build GRCh38
rs780893434 Chr.12: 102402514-102402514 on Build GRCh38
rs766840468 Chr.12: 102402614-102402614 on Build GRCh38
rs371947838 Chr.12: 102402244-102402245 on Build GRCh38
rs762745129 Chr.12: 102402395-102402395 on Build GRCh38
rs191847837 Chr.12: 102402678-102402678 on Build GRCh38
rs374864970 Chr.12: 102402484-102402484 on Build GRCh38
rs199909639 Chr.12: 102402501-102402501 on Build GRCh38
rs993267852 Chr.12: 102402249-102402249 on Build GRCh38
rs539494367 Chr.12: 102402244-102402244 on Build GRCh38
rs536666389 Chr.12: 102402549-102402549 on Build GRCh38
rs78177141 Chr.12: 102402384-102402385 on Build GRCh38
rs765479439 Chr.12: 102402607-102402607 on Build GRCh38
rs980869244 Chr.12: 102402312-102402312 on Build GRCh38
rs749864229 Chr.12: 102402658-102402658 on Build GRCh38
rs3032449 Chr.12: 102402246-102402246 on Build GRCh38
rs565255968 Chr.12: 102402682-102402682 on Build GRCh38
rs5742698 Chr.12: 102402480-102402480 on Build GRCh38
rs550911517 Chr.12: 102402367-102402367 on Build GRCh38
rs765532484 Chr.12: 102402473-102402473 on Build GRCh38
rs5031032 Chr.12: 102402354-102402354 on Build GRCh38
rs958954985 Chr.12: 102402346-102402346 on Build GRCh38
rs930732713 Chr.12: 102402659-102402659 on Build GRCh38
rs149540080 Chr.12: 102402402-102402402 on Build GRCh38
rs756839832 Chr.12: 102402229-102402230 on Build GRCh38
rs367987264 Chr.12: 102402470-102402470 on Build GRCh38
rs930861486 Chr.12: 102402379-102402379 on Build GRCh38
rs565327837 Chr.12: 102402248-102402248 on Build GRCh38
rs775506745 Chr.12: 102402336-102402336 on Build GRCh38
rs763358713 Chr.12: 102402610-102402610 on Build GRCh38
rs1045410736 Chr.12: 102402442-102402442 on Build GRCh38
rs757012625 Chr.12: 102402456-102402456 on Build GRCh38
rs530300044 Chr.12: 102402469-102402469 on Build GRCh38
rs530814211 Chr.12: 102402262-102402262 on Build GRCh38
rs1047896501 Chr.12: 102402686-102402686 on Build GRCh38
rs70961704 Chr.12: 102402244-102402244 on Build GRCh38
rs1039142847 Chr.12: 102402338-102402338 on Build GRCh38
rs535492231 Chr.12: 102402691-102402691 on Build GRCh38
rs777643169 Chr.12: 102402531-102402531 on Build GRCh38
rs1050870079 Chr.12: 102402422-102402422 on Build GRCh38
rs776789001 Chr.12: 102402591-102402591 on Build GRCh38
rs771502062 Chr.12: 102402242-102402242 on Build GRCh38
rs769499253 Chr.12: 102402529-102402529 on Build GRCh38
rs886667040 Chr.12: 102402385-102402385 on Build GRCh38
rs754487328 Chr.12: 102402510-102402510 on Build GRCh38
rs115707403 Chr.12: 102402507-102402507 on Build GRCh38
rs964781817 Chr.12: 102402613-102402613 on Build GRCh38
rs747812688 Chr.12: 102402519-102402519 on Build GRCh38
rs61250514 Chr.12: 102402384-102402384 on Build GRCh38
rs767878440 Chr.12: 102402228-102402229 on Build GRCh38
rs955265175 Chr.12: 102402493-102402493 on Build GRCh38
rs751113096 Chr.12: 102402449-102402449 on Build GRCh38
rs889009661 Chr.12: 102402448-102402448 on Build GRCh38
rs780224631 Chr.12: 102402648-102402648 on Build GRCh38
rs911907364 Chr.12: 102402297-102402297 on Build GRCh38
rs559568729 Chr.12: 102402220-102402220 on Build GRCh38
rs750741271 Chr.12: 102402608-102402608 on Build GRCh38
rs757634465 Chr.12: 102402472-102402472 on Build GRCh38
rs201258075 Chr.12: 102402657-102402657 on Build GRCh38
rs761877010 Chr.12: 102402599-102402599 on Build GRCh38
rs951742304 Chr.12: 102402291-102402291 on Build GRCh38
rs199891259 Chr.12: 102402246-102402247 on Build GRCh38
rs367603396 Chr.12: 102402548-102402548 on Build GRCh38
rs756533031 Chr.12: 102402522-102402522 on Build GRCh38
rs768682086 Chr.12: 102402585-102402585 on Build GRCh38
rs760690455 Chr.12: 102402467-102402467 on Build GRCh38
rs879384585 Chr.12: 102402247-102402247 on Build GRCh38
rs1001687302 Chr.12: 102402452-102402452 on Build GRCh38
rs749124997 Chr.12: 102402539-102402539 on Build GRCh38
rs943437019 Chr.12: 102402302-102402302 on Build GRCh38
rs747667102 Chr.12: 102402574-102402575 on Build GRCh38
rs920759124 Chr.12: 102402373-102402373 on Build GRCh38
rs574772628 Chr.12: 102402660-102402660 on Build GRCh38
rs886048889 Chr.12: 102402242-102402244 on Build GRCh38
rs187371655 Chr.12: 102402476-102402476 on Build GRCh38
rs780619080 Chr.12: 102402569-102402572 on Build GRCh38
rs967521565 Chr.12: 102402699-102402699 on Build GRCh38
rs14970 Chr.12: 102402401-102402401 on Build GRCh38
rs975224671 Chr.12: 102402631-102402631 on Build GRCh38

View Full Product Details