Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21670993-21671489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF9
Entrez Gene ID
2254
Gene Name
fibroblast growth factor 9
Gene Aliases
FGF-9, GAF, HBFG-9, HBGF-9, SYNS3
UniGene
Hs.111
Gene Chromosome Location
Chr.13: 21671076-21704501 on Build GRCh38
Transcripts
NM_002010.2

SNP Detail

SNP ID Chromosome Location
rs948498332 Chr.13: 21671337-21671337 on Build GRCh38
rs1045520813 Chr.13: 21671281-21671281 on Build GRCh38
rs924691588 Chr.13: 21671134-21671134 on Build GRCh38
rs559627485 Chr.13: 21671114-21671114 on Build GRCh38
rs796479555 Chr.13: 21671146-21671146 on Build GRCh38
rs976087919 Chr.13: 21671099-21671099 on Build GRCh38
rs753940318 Chr.13: 21671356-21671356 on Build GRCh38
rs971345564 Chr.13: 21671032-21671032 on Build GRCh38
rs374877065 Chr.13: 21671333-21671333 on Build GRCh38
rs953543815 Chr.13: 21671198-21671198 on Build GRCh38
rs916733919 Chr.13: 21671251-21671251 on Build GRCh38
rs750719377 Chr.13: 21671056-21671056 on Build GRCh38
rs923422721 Chr.13: 21671091-21671091 on Build GRCh38
rs538791227 Chr.13: 21671221-21671221 on Build GRCh38
rs547446043 Chr.13: 21671246-21671246 on Build GRCh38
rs964730413 Chr.13: 21671091-21671091 on Build GRCh38
rs886050041 Chr.13: 21671461-21671461 on Build GRCh38
rs900839325 Chr.13: 21671465-21671465 on Build GRCh38
rs1036727709 Chr.13: 21671417-21671417 on Build GRCh38
rs1012670935 Chr.13: 21671140-21671140 on Build GRCh38
rs983446612 Chr.13: 21671204-21671204 on Build GRCh38
rs923301415 Chr.13: 21671199-21671199 on Build GRCh38
rs1017428199 Chr.13: 21671186-21671186 on Build GRCh38
rs1055266951 Chr.13: 21671131-21671131 on Build GRCh38
rs906929212 Chr.13: 21671286-21671286 on Build GRCh38
rs565936638 Chr.13: 21671385-21671385 on Build GRCh38
rs989976452 Chr.13: 21671086-21671086 on Build GRCh38
rs531745333 Chr.13: 21671187-21671187 on Build GRCh38
rs1012874638 Chr.13: 21671141-21671141 on Build GRCh38
rs938779165 Chr.13: 21671247-21671247 on Build GRCh38
rs563405759 Chr.13: 21671139-21671139 on Build GRCh38
rs942210347 Chr.13: 21671129-21671129 on Build GRCh38
rs576891863 Chr.13: 21671083-21671083 on Build GRCh38
rs1048431691 Chr.13: 21671119-21671119 on Build GRCh38
rs909436329 Chr.13: 21671124-21671124 on Build GRCh38
rs998756155 Chr.13: 21671052-21671052 on Build GRCh38
rs549054241 Chr.13: 21671117-21671117 on Build GRCh38
rs1015307391 Chr.13: 21671057-21671057 on Build GRCh38
rs976119197 Chr.13: 21671190-21671190 on Build GRCh38
rs1011831301 Chr.13: 21671181-21671181 on Build GRCh38
rs370588961 Chr.13: 21671210-21671210 on Build GRCh38
rs915787981 Chr.13: 21671322-21671323 on Build GRCh38
rs1057013997 Chr.13: 21671137-21671137 on Build GRCh38
rs888345532 Chr.13: 21671375-21671376 on Build GRCh38
rs537581205 Chr.13: 21671138-21671138 on Build GRCh38
rs910630936 Chr.13: 21671088-21671088 on Build GRCh38
rs557866920 Chr.13: 21671445-21671445 on Build GRCh38
rs1010410671 Chr.13: 21671135-21671135 on Build GRCh38
rs536421616 Chr.13: 21671387-21671387 on Build GRCh38
rs965178830 Chr.13: 21671188-21671188 on Build GRCh38
rs914596262 Chr.13: 21671105-21671105 on Build GRCh38
rs998931240 Chr.13: 21671340-21671340 on Build GRCh38
rs376388740 Chr.13: 21671201-21671201 on Build GRCh38
rs757348372 Chr.13: 21671453-21671453 on Build GRCh38
rs1045450611 Chr.13: 21671142-21671142 on Build GRCh38
rs906854265 Chr.13: 21671145-21671145 on Build GRCh38
rs962533023 Chr.13: 21671072-21671072 on Build GRCh38
rs942290748 Chr.13: 21671212-21671212 on Build GRCh38
rs543016860 Chr.13: 21671023-21671023 on Build GRCh38
rs1048801271 Chr.13: 21671369-21671369 on Build GRCh38
rs567350418 Chr.13: 21671136-21671136 on Build GRCh38
rs555608414 Chr.13: 21671307-21671308 on Build GRCh38
rs1031532486 Chr.13: 21671054-21671054 on Build GRCh38
rs541124749 Chr.13: 21671104-21671104 on Build GRCh38
rs977425496 Chr.13: 21671224-21671224 on Build GRCh38
rs886050040 Chr.13: 21671328-21671329 on Build GRCh38
rs1003853233 Chr.13: 21671168-21671168 on Build GRCh38
rs564880858 Chr.13: 21671081-21671081 on Build GRCh38
rs948483531 Chr.13: 21671267-21671267 on Build GRCh38
rs1032025635 Chr.13: 21671173-21671173 on Build GRCh38
rs888116599 Chr.13: 21671163-21671163 on Build GRCh38
rs957272445 Chr.13: 21671175-21671175 on Build GRCh38
rs1042202915 Chr.13: 21671122-21671122 on Build GRCh38

View Full Product Details