Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21700942-21701443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF9
Entrez Gene ID
2254
Gene Name
fibroblast growth factor 9
Gene Aliases
FGF-9, GAF, HBFG-9, HBGF-9, SYNS3
UniGene
Hs.111
Gene Chromosome Location
Chr.13: 21671076-21704501 on Build GRCh38
Transcripts
NM_002010.2 XM_011534996.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7093276 c.499A>G p.N167D Chr.13: 21701307-21701307 on Build GRCh38
1706514 c.520G>A p.E174K Chr.13: 21701328-21701328 on Build GRCh38
4962730 c.622A>C p.S208R Chr.13: 21701430-21701430 on Build GRCh38
1716987 c.603T>C p.Y201Y Chr.13: 21701411-21701411 on Build GRCh38
5132866 c.421G>A p.E141K Chr.13: 21701229-21701229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747992576 Chr.13: 21701179-21701179 on Build GRCh38
rs550814848 Chr.13: 21701347-21701347 on Build GRCh38
rs143118647 Chr.13: 21701273-21701273 on Build GRCh38
rs771825272 Chr.13: 21701180-21701180 on Build GRCh38
rs746006892 Chr.13: 21701111-21701111 on Build GRCh38
rs780247811 Chr.13: 21701174-21701174 on Build GRCh38
rs765041263 Chr.13: 21701228-21701228 on Build GRCh38
rs764396440 Chr.13: 21701249-21701249 on Build GRCh38
rs1037215966 Chr.13: 21701093-21701093 on Build GRCh38
rs865865517 Chr.13: 21701387-21701387 on Build GRCh38
rs9509841 Chr.13: 21701255-21701255 on Build GRCh38
rs866384101 Chr.13: 21701384-21701384 on Build GRCh38
rs997316078 Chr.13: 21701143-21701143 on Build GRCh38
rs17840925 Chr.13: 21701062-21701062 on Build GRCh38
rs267603779 Chr.13: 21701411-21701411 on Build GRCh38
rs762716906 Chr.13: 21701330-21701330 on Build GRCh38
rs369272027 Chr.13: 21701391-21701391 on Build GRCh38
rs1030487041 Chr.13: 21701146-21701146 on Build GRCh38
rs267603778 Chr.13: 21701313-21701313 on Build GRCh38
rs519234 Chr.13: 21701071-21701071 on Build GRCh38
rs200015821 Chr.13: 21701208-21701208 on Build GRCh38
rs770003715 Chr.13: 21701172-21701172 on Build GRCh38
rs146697820 Chr.13: 21701290-21701290 on Build GRCh38
rs371548027 Chr.13: 21701323-21701323 on Build GRCh38
rs647236 Chr.13: 21700988-21700988 on Build GRCh38
rs781152657 Chr.13: 21701166-21701166 on Build GRCh38
rs916299225 Chr.13: 21701037-21701037 on Build GRCh38
rs868277032 Chr.13: 21701415-21701415 on Build GRCh38
rs950414702 Chr.13: 21701150-21701150 on Build GRCh38
rs527354619 Chr.13: 21701321-21701321 on Build GRCh38
rs575142925 Chr.13: 21701013-21701013 on Build GRCh38
rs751978201 Chr.13: 21701164-21701164 on Build GRCh38
rs540047092 Chr.13: 21701126-21701126 on Build GRCh38
rs779192127 Chr.13: 21701399-21701399 on Build GRCh38
rs538431086 Chr.13: 21701032-21701032 on Build GRCh38
rs963305629 Chr.13: 21701248-21701248 on Build GRCh38
rs746838387 Chr.13: 21701402-21701402 on Build GRCh38
rs745885877 Chr.13: 21701170-21701170 on Build GRCh38
rs1035113584 Chr.13: 21701005-21701005 on Build GRCh38
rs518544 Chr.13: 21701054-21701054 on Build GRCh38
rs759113908 Chr.13: 21701216-21701216 on Build GRCh38
rs746751637 Chr.13: 21701308-21701308 on Build GRCh38
rs749546597 Chr.13: 21701274-21701274 on Build GRCh38
rs770847282 Chr.13: 21701207-21701207 on Build GRCh38
rs778392064 Chr.13: 21701160-21701160 on Build GRCh38
rs1010781871 Chr.13: 21701229-21701229 on Build GRCh38
rs267603777 Chr.13: 21701202-21701202 on Build GRCh38
rs902550368 Chr.13: 21701018-21701018 on Build GRCh38
rs762229622 Chr.13: 21701353-21701353 on Build GRCh38
rs1022253383 Chr.13: 21700976-21700976 on Build GRCh38
rs201876493 Chr.13: 21701324-21701324 on Build GRCh38
rs767964377 Chr.13: 21701360-21701360 on Build GRCh38
rs753392097 Chr.13: 21701063-21701063 on Build GRCh38
rs756170423 Chr.13: 21701270-21701270 on Build GRCh38
rs751903634 Chr.13: 21701254-21701254 on Build GRCh38
rs750565955 Chr.13: 21701262-21701262 on Build GRCh38
rs562768508 Chr.13: 21700997-21700997 on Build GRCh38
rs776618974 Chr.13: 21701322-21701322 on Build GRCh38
rs750904852 Chr.13: 21701390-21701390 on Build GRCh38
rs755164853 Chr.13: 21701392-21701392 on Build GRCh38
rs984252256 Chr.13: 21701061-21701061 on Build GRCh38
rs960380409 Chr.13: 21701011-21701011 on Build GRCh38
rs763743837 Chr.13: 21701346-21701346 on Build GRCh38
rs933967044 Chr.13: 21701171-21701171 on Build GRCh38
rs758824655 Chr.13: 21701139-21701139 on Build GRCh38
rs1007072429 Chr.13: 21701076-21701076 on Build GRCh38
rs935325287 Chr.13: 21701019-21701019 on Build GRCh38
rs879396163 Chr.13: 21701044-21701044 on Build GRCh38
rs187308315 Chr.13: 21701231-21701231 on Build GRCh38
rs773022513 Chr.13: 21701184-21701184 on Build GRCh38
rs1046897405 Chr.13: 21701012-21701012 on Build GRCh38

View Full Product Details