Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21702393-21702906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF9
Entrez Gene ID
2254
Gene Name
fibroblast growth factor 9
Gene Aliases
FGF-9, GAF, HBFG-9, HBGF-9, SYNS3
UniGene
Hs.111
Gene Chromosome Location
Chr.13: 21671076-21704501 on Build GRCh38
Transcripts
NM_002010.2 XM_011534996.2

SNP Detail

SNP ID Chromosome Location
rs776882017 Chr.13: 21702419-21702419 on Build GRCh38
rs980568819 Chr.13: 21702654-21702654 on Build GRCh38
rs956898361 Chr.13: 21702625-21702625 on Build GRCh38
rs555464634 Chr.13: 21702639-21702639 on Build GRCh38
rs962967285 Chr.13: 21702584-21702584 on Build GRCh38
rs528529449 Chr.13: 21702827-21702827 on Build GRCh38
rs370206395 Chr.13: 21702701-21702701 on Build GRCh38
rs529908179 Chr.13: 21702539-21702539 on Build GRCh38
rs373802130 Chr.13: 21702706-21702706 on Build GRCh38
rs956739510 Chr.13: 21702829-21702829 on Build GRCh38
rs946661451 Chr.13: 21702736-21702736 on Build GRCh38
rs578002791 Chr.13: 21702738-21702738 on Build GRCh38
rs1025095632 Chr.13: 21702838-21702838 on Build GRCh38
rs899700183 Chr.13: 21702780-21702780 on Build GRCh38
rs1015578105 Chr.13: 21702545-21702545 on Build GRCh38
rs992257487 Chr.13: 21702629-21702629 on Build GRCh38
rs17070748 Chr.13: 21702431-21702431 on Build GRCh38
rs551156731 Chr.13: 21702865-21702865 on Build GRCh38
rs562504800 Chr.13: 21702713-21702713 on Build GRCh38
rs1000898581 Chr.13: 21702621-21702621 on Build GRCh38
rs1043571552 Chr.13: 21702425-21702425 on Build GRCh38
rs573648710 Chr.13: 21702674-21702674 on Build GRCh38
rs537888875 Chr.13: 21702660-21702660 on Build GRCh38
rs891236680 Chr.13: 21702814-21702814 on Build GRCh38
rs997285391 Chr.13: 21702492-21702492 on Build GRCh38
rs772193978 Chr.13: 21702467-21702467 on Build GRCh38
rs917960237 Chr.13: 21702638-21702638 on Build GRCh38
rs1043647823 Chr.13: 21702768-21702768 on Build GRCh38
rs927738757 Chr.13: 21702663-21702663 on Build GRCh38
rs545298237 Chr.13: 21702745-21702745 on Build GRCh38
rs962686468 Chr.13: 21702843-21702843 on Build GRCh38
rs939081515 Chr.13: 21702673-21702673 on Build GRCh38
rs1051557769 Chr.13: 21702762-21702762 on Build GRCh38
rs996696786 Chr.13: 21702781-21702781 on Build GRCh38
rs536710049 Chr.13: 21702730-21702730 on Build GRCh38
rs963155825 Chr.13: 21702856-21702856 on Build GRCh38
rs1031022181 Chr.13: 21702624-21702625 on Build GRCh38
rs974308204 Chr.13: 21702606-21702606 on Build GRCh38
rs1039883672 Chr.13: 21702801-21702801 on Build GRCh38
rs1029741781 Chr.13: 21702460-21702460 on Build GRCh38
rs747536753 Chr.13: 21702790-21702790 on Build GRCh38
rs529361203 Chr.13: 21702852-21702852 on Build GRCh38
rs969858348 Chr.13: 21702845-21702845 on Build GRCh38
rs553726872 Chr.13: 21702445-21702445 on Build GRCh38
rs970676773 Chr.13: 21702881-21702881 on Build GRCh38
rs761863548 Chr.13: 21702493-21702493 on Build GRCh38
rs564490885 Chr.13: 21702795-21702795 on Build GRCh38
rs891305046 Chr.13: 21702519-21702519 on Build GRCh38
rs181623879 Chr.13: 21702846-21702846 on Build GRCh38
rs546656888 Chr.13: 21702828-21702828 on Build GRCh38
rs903994231 Chr.13: 21702809-21702809 on Build GRCh38
rs3715 Chr.13: 21702529-21702529 on Build GRCh38
rs909155278 Chr.13: 21702656-21702656 on Build GRCh38
rs932948820 Chr.13: 21702727-21702727 on Build GRCh38
rs916086922 Chr.13: 21702618-21702618 on Build GRCh38
rs576528151 Chr.13: 21702848-21702848 on Build GRCh38
rs1013995250 Chr.13: 21702836-21702836 on Build GRCh38

View Full Product Details