Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 61679-62169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBDF1
Entrez Gene ID
64285
Gene Name
rhomboid 5 homolog 1
Gene Aliases
C16orf8, Dist1, EGFR-RS, gene-89, gene-90, hDist1
UniGene
Hs.744992
Gene Chromosome Location
Chr.16: 58059-76355 on Build GRCh38
Transcripts
NM_022450.3 XM_005255494.1 XM_005255498.2 XM_006720921.1 XM_017023556.1 XM_017023557.1 XM_017023558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4748241 c.1143C>A p.N381K Chr.16: 61863-61863 on Build GRCh38
966970 c.1174G>A p.V392I Chr.16: 61832-61832 on Build GRCh38
1223685 c.1060C>T p.Q354* Chr.16: 61946-61946 on Build GRCh38
6766088 c.1049C>T p.P350L Chr.16: 61957-61957 on Build GRCh38
6766077 c.1064G>A p.R355H Chr.16: 61942-61942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030366397 Chr.16: 61919-61919 on Build GRCh38
rs770120571 Chr.16: 62033-62033 on Build GRCh38
rs201123556 Chr.16: 61962-61962 on Build GRCh38
rs577546342 Chr.16: 62078-62078 on Build GRCh38
rs202235681 Chr.16: 61738-61738 on Build GRCh38
rs775568789 Chr.16: 61756-61756 on Build GRCh38
rs757076733 Chr.16: 61963-61963 on Build GRCh38
rs750444515 Chr.16: 61984-61984 on Build GRCh38
rs369432381 Chr.16: 61820-61820 on Build GRCh38
rs998763769 Chr.16: 62136-62136 on Build GRCh38
rs773590799 Chr.16: 61899-61899 on Build GRCh38
rs558178061 Chr.16: 61887-61887 on Build GRCh38
rs772249847 Chr.16: 61951-61951 on Build GRCh38
rs1030581901 Chr.16: 61719-61719 on Build GRCh38
rs935065111 Chr.16: 62027-62027 on Build GRCh38
rs749421040 Chr.16: 61839-61839 on Build GRCh38
rs770971284 Chr.16: 62016-62016 on Build GRCh38
rs755971252 Chr.16: 61987-61987 on Build GRCh38
rs761566719 Chr.16: 61834-61834 on Build GRCh38
rs534256998 Chr.16: 61933-61933 on Build GRCh38
rs757887168 Chr.16: 61707-61707 on Build GRCh38
rs777379972 Chr.16: 61925-61925 on Build GRCh38
rs747121791 Chr.16: 62015-62015 on Build GRCh38
rs768944027 Chr.16: 61938-61938 on Build GRCh38
rs762370207 Chr.16: 61768-61768 on Build GRCh38
rs974315128 Chr.16: 61805-61805 on Build GRCh38
rs779265495 Chr.16: 61943-61943 on Build GRCh38
rs963515127 Chr.16: 61989-61989 on Build GRCh38
rs752314587 Chr.16: 61959-61959 on Build GRCh38
rs918923585 Chr.16: 62024-62024 on Build GRCh38
rs763276496 Chr.16: 61759-61759 on Build GRCh38
rs916852082 Chr.16: 61721-61721 on Build GRCh38
rs368502758 Chr.16: 61704-61704 on Build GRCh38
rs779588125 Chr.16: 61729-61729 on Build GRCh38
rs745662656 Chr.16: 61937-61937 on Build GRCh38
rs915012611 Chr.16: 62076-62076 on Build GRCh38
rs954724760 Chr.16: 61979-61979 on Build GRCh38
rs774595922 Chr.16: 61764-61764 on Build GRCh38
rs749030496 Chr.16: 61736-61736 on Build GRCh38
rs544020672 Chr.16: 62019-62019 on Build GRCh38
rs185723860 Chr.16: 61785-61785 on Build GRCh38
rs762641834 Chr.16: 61961-61961 on Build GRCh38
rs760075059 Chr.16: 62023-62023 on Build GRCh38
rs778438223 Chr.16: 61741-61741 on Build GRCh38
rs188922031 Chr.16: 62133-62133 on Build GRCh38
rs764880997 Chr.16: 61916-61916 on Build GRCh38
rs773010448 Chr.16: 61700-61700 on Build GRCh38
rs73493080 Chr.16: 62081-62081 on Build GRCh38
rs374656396 Chr.16: 61890-61890 on Build GRCh38
rs777679119 Chr.16: 62014-62014 on Build GRCh38
rs749223491 Chr.16: 62022-62024 on Build GRCh38
rs527299346 Chr.16: 62089-62089 on Build GRCh38
rs764246154 Chr.16: 61760-61760 on Build GRCh38
rs923652451 Chr.16: 62022-62022 on Build GRCh38
rs766634557 Chr.16: 61910-61910 on Build GRCh38
rs199962084 Chr.16: 61703-61703 on Build GRCh38
rs773394842 Chr.16: 61952-61952 on Build GRCh38
rs761373878 Chr.16: 62075-62075 on Build GRCh38
rs539820234 Chr.16: 61789-61789 on Build GRCh38
rs368233422 Chr.16: 61749-61749 on Build GRCh38
rs560987617 Chr.16: 62118-62118 on Build GRCh38
rs755726797 Chr.16: 61716-61716 on Build GRCh38
rs748451457 Chr.16: 61945-61945 on Build GRCh38
rs372098045 Chr.16: 61782-61782 on Build GRCh38
rs1054011240 Chr.16: 61757-61757 on Build GRCh38
rs775854063 Chr.16: 62038-62038 on Build GRCh38
rs750785112 Chr.16: 61711-61711 on Build GRCh38
rs1052143485 Chr.16: 62143-62143 on Build GRCh38
rs77201493 Chr.16: 61956-61956 on Build GRCh38
rs761287711 Chr.16: 61909-61909 on Build GRCh38
rs774505993 Chr.16: 61864-61864 on Build GRCh38
rs751429564 Chr.16: 61929-61929 on Build GRCh38
rs752402341 Chr.16: 61917-61917 on Build GRCh38
rs216601 Chr.16: 62114-62114 on Build GRCh38
rs371815212 Chr.16: 62004-62004 on Build GRCh38
rs990987836 Chr.16: 61998-61998 on Build GRCh38
rs772023662 Chr.16: 61745-61745 on Build GRCh38
rs763812227 Chr.16: 62056-62056 on Build GRCh38
rs777672313 Chr.16: 61811-61811 on Build GRCh38
rs758272706 Chr.16: 61803-61803 on Build GRCh38
rs938878513 Chr.16: 62063-62063 on Build GRCh38
rs765379485 Chr.16: 61701-61701 on Build GRCh38
rs778351952 Chr.16: 61792-61792 on Build GRCh38
rs747657518 Chr.16: 61795-61795 on Build GRCh38
rs769032030 Chr.16: 61847-61847 on Build GRCh38
rs947476943 Chr.16: 62091-62091 on Build GRCh38
rs755876138 Chr.16: 61706-61707 on Build GRCh38
rs781202738 Chr.16: 61966-61966 on Build GRCh38
rs562429629 Chr.16: 62043-62043 on Build GRCh38
rs772365601 Chr.16: 62013-62013 on Build GRCh38
rs757898679 Chr.16: 61924-61924 on Build GRCh38
rs747033872 Chr.16: 61818-61818 on Build GRCh38
rs750743703 Chr.16: 61779-61779 on Build GRCh38
rs770816376 Chr.16: 61755-61755 on Build GRCh38
rs544678216 Chr.16: 62079-62079 on Build GRCh38
rs777620317 Chr.16: 61991-61991 on Build GRCh38
rs773678451 Chr.16: 62065-62065 on Build GRCh38
rs376370477 Chr.16: 61773-61773 on Build GRCh38
rs536059853 Chr.16: 61709-61709 on Build GRCh38
rs748361604 Chr.16: 62007-62007 on Build GRCh38
rs759118513 Chr.16: 61913-61913 on Build GRCh38
rs780810982 Chr.16: 61936-61936 on Build GRCh38
rs780037413 Chr.16: 62005-62005 on Build GRCh38
rs930226218 Chr.16: 61842-61842 on Build GRCh38
rs759878923 Chr.16: 61957-61957 on Build GRCh38
rs150936366 Chr.16: 61912-61912 on Build GRCh38
rs746791119 Chr.16: 61753-61753 on Build GRCh38

View Full Product Details