Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3725030-3725531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
UniGene
Hs.459759
Gene Chromosome Location
Chr.16: 3725054-3880727 on Build GRCh38
Transcripts
NM_001079846.1 NM_004380.2 XM_005255124.4 XM_005255125.4 XM_006720848.3 XM_011522381.2 XM_017022944.1

SNP Detail

SNP ID Chromosome Location
rs975485103 Chr.16: 3725451-3725451 on Build GRCh38
rs375742594 Chr.16: 3725220-3725220 on Build GRCh38
rs988554666 Chr.16: 3725259-3725259 on Build GRCh38
rs1044154828 Chr.16: 3725362-3725362 on Build GRCh38
rs142823437 Chr.16: 3725444-3725444 on Build GRCh38
rs908767259 Chr.16: 3725481-3725481 on Build GRCh38
rs1003338389 Chr.16: 3725354-3725354 on Build GRCh38
rs1054695484 Chr.16: 3725484-3725484 on Build GRCh38
rs760630174 Chr.16: 3725222-3725222 on Build GRCh38
rs193262955 Chr.16: 3725251-3725251 on Build GRCh38
rs542425636 Chr.16: 3725483-3725483 on Build GRCh38
rs922704211 Chr.16: 3725285-3725285 on Build GRCh38
rs531651621 Chr.16: 3725095-3725095 on Build GRCh38
rs959252592 Chr.16: 3725372-3725372 on Build GRCh38
rs567048198 Chr.16: 3725244-3725244 on Build GRCh38
rs567712178 Chr.16: 3725154-3725154 on Build GRCh38
rs766344592 Chr.16: 3725315-3725315 on Build GRCh38
rs984020080 Chr.16: 3725410-3725410 on Build GRCh38
rs538684283 Chr.16: 3725332-3725332 on Build GRCh38
rs934169696 Chr.16: 3725305-3725305 on Build GRCh38
rs1031585110 Chr.16: 3725226-3725226 on Build GRCh38
rs189649723 Chr.16: 3725176-3725176 on Build GRCh38
rs890004855 Chr.16: 3725318-3725318 on Build GRCh38
rs975516163 Chr.16: 3725298-3725298 on Build GRCh38
rs911407197 Chr.16: 3725455-3725455 on Build GRCh38
rs1033481526 Chr.16: 3725364-3725364 on Build GRCh38
rs761667070 Chr.16: 3725191-3725191 on Build GRCh38
rs926539117 Chr.16: 3725221-3725221 on Build GRCh38
rs959283878 Chr.16: 3725233-3725233 on Build GRCh38
rs1053422306 Chr.16: 3725328-3725328 on Build GRCh38
rs566521450 Chr.16: 3725331-3725331 on Build GRCh38
rs9392 Chr.16: 3725168-3725168 on Build GRCh38
rs898223973 Chr.16: 3725348-3725348 on Build GRCh38
rs184069497 Chr.16: 3725453-3725453 on Build GRCh38
rs961386426 Chr.16: 3725429-3725429 on Build GRCh38
rs573633262 Chr.16: 3725495-3725495 on Build GRCh38
rs948424452 Chr.16: 3725507-3725507 on Build GRCh38
rs182274996 Chr.16: 3725180-3725180 on Build GRCh38
rs569415301 Chr.16: 3725263-3725263 on Build GRCh38
rs956163210 Chr.16: 3725443-3725443 on Build GRCh38
rs886768121 Chr.16: 3725149-3725151 on Build GRCh38
rs117551000 Chr.16: 3725351-3725351 on Build GRCh38
rs900059419 Chr.16: 3725094-3725094 on Build GRCh38
rs570738685 Chr.16: 3725148-3725152 on Build GRCh38
rs771949537 Chr.16: 3725213-3725213 on Build GRCh38
rs538167515 Chr.16: 3725323-3725323 on Build GRCh38
rs986976652 Chr.16: 3725469-3725469 on Build GRCh38
rs550968281 Chr.16: 3725118-3725118 on Build GRCh38
rs375372620 Chr.16: 3725089-3725089 on Build GRCh38
rs569468762 Chr.16: 3725438-3725438 on Build GRCh38
rs957742172 Chr.16: 3725173-3725173 on Build GRCh38
rs558800860 Chr.16: 3725432-3725432 on Build GRCh38
rs1000458775 Chr.16: 3725326-3725326 on Build GRCh38
rs893495968 Chr.16: 3725329-3725329 on Build GRCh38
rs535625078 Chr.16: 3725250-3725250 on Build GRCh38
rs1037098827 Chr.16: 3725266-3725266 on Build GRCh38
rs764200235 Chr.16: 3725344-3725344 on Build GRCh38
rs748838577 Chr.16: 3725065-3725065 on Build GRCh38
rs968103256 Chr.16: 3725449-3725449 on Build GRCh38
rs187552100 Chr.16: 3725241-3725241 on Build GRCh38
rs1016827752 Chr.16: 3725414-3725414 on Build GRCh38
rs907037889 Chr.16: 3725389-3725389 on Build GRCh38
rs188658312 Chr.16: 3725493-3725493 on Build GRCh38
rs940073959 Chr.16: 3725260-3725260 on Build GRCh38
rs546740096 Chr.16: 3725201-3725201 on Build GRCh38
rs752714303 Chr.16: 3725316-3725316 on Build GRCh38
rs539938276 Chr.16: 3725425-3725425 on Build GRCh38

View Full Product Details