Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31909689-31910208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTP4A2
Entrez Gene ID
8073
Gene Name
protein tyrosine phosphatase type IVA, member 2
Gene Aliases
HH13, HH7-2, HU-PP-1, OV-1, PRL-2, PRL2, PTP4A, PTPCAAX2, ptp-IV1a, ptp-IV1b
UniGene
Hs.470477
Gene Chromosome Location
Chr.1: 31906421-31942635 on Build GRCh38
Transcripts
NM_001195100.1 NM_001195101.1 NM_080391.3 XM_005271231.3 XM_005271232.3 XM_017002401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1222501 c.323C>T p.A108V Chr.1: 31910110-31910110 on Build GRCh38
7312250 c.369C>A p.Y123* Chr.1: 31910064-31910064 on Build GRCh38
4577221 c.339A>G p.A113A Chr.1: 31910094-31910094 on Build GRCh38
6758771 c.357T>C p.C119C Chr.1: 31910076-31910076 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs926710339 Chr.1: 31909908-31909908 on Build GRCh38
rs538578470 Chr.1: 31910176-31910176 on Build GRCh38
rs149063246 Chr.1: 31909762-31909762 on Build GRCh38
rs12094379 Chr.1: 31910145-31910145 on Build GRCh38
rs377321954 Chr.1: 31910021-31910021 on Build GRCh38
rs575387964 Chr.1: 31909855-31909855 on Build GRCh38
rs997237529 Chr.1: 31909747-31909747 on Build GRCh38
rs936906335 Chr.1: 31909776-31909776 on Build GRCh38
rs1053961516 Chr.1: 31909800-31909800 on Build GRCh38
rs528297246 Chr.1: 31909877-31909877 on Build GRCh38
rs1055623833 Chr.1: 31909953-31909953 on Build GRCh38
rs550698067 Chr.1: 31909969-31909969 on Build GRCh38
rs568941180 Chr.1: 31910025-31910025 on Build GRCh38
rs996497107 Chr.1: 31909875-31909875 on Build GRCh38
rs972522297 Chr.1: 31909720-31909720 on Build GRCh38
rs952667792 Chr.1: 31909900-31909900 on Build GRCh38
rs756683728 Chr.1: 31910033-31910033 on Build GRCh38
rs145123194 Chr.1: 31909742-31909742 on Build GRCh38
rs545954433 Chr.1: 31909866-31909866 on Build GRCh38
rs753328143 Chr.1: 31910077-31910077 on Build GRCh38
rs372009429 Chr.1: 31909977-31909977 on Build GRCh38
rs779795895 Chr.1: 31910157-31910157 on Build GRCh38
rs972043130 Chr.1: 31909878-31909878 on Build GRCh38
rs749491395 Chr.1: 31909766-31909766 on Build GRCh38
rs1007711572 Chr.1: 31909781-31909781 on Build GRCh38
rs982066187 Chr.1: 31910170-31910170 on Build GRCh38
rs561844679 Chr.1: 31909898-31909898 on Build GRCh38
rs141172283 Chr.1: 31910064-31910064 on Build GRCh38
rs369365506 Chr.1: 31910002-31910002 on Build GRCh38
rs564239417 Chr.1: 31909872-31909872 on Build GRCh38
rs918035922 Chr.1: 31909996-31909996 on Build GRCh38
rs778584358 Chr.1: 31910133-31910133 on Build GRCh38
rs984403257 Chr.1: 31909907-31909907 on Build GRCh38
rs893041382 Chr.1: 31909718-31909718 on Build GRCh38
rs534146937 Chr.1: 31910160-31910160 on Build GRCh38
rs529915055 Chr.1: 31910169-31910169 on Build GRCh38
rs765927878 Chr.1: 31909889-31909892 on Build GRCh38
rs914513533 Chr.1: 31909961-31909961 on Build GRCh38
rs937202111 Chr.1: 31909923-31909923 on Build GRCh38
rs768518685 Chr.1: 31909993-31909993 on Build GRCh38
rs574231495 Chr.1: 31910165-31910165 on Build GRCh38
rs771185220 Chr.1: 31909848-31909848 on Build GRCh38
rs982139869 Chr.1: 31909896-31909896 on Build GRCh38
rs146841828 Chr.1: 31909755-31909755 on Build GRCh38
rs900911477 Chr.1: 31909869-31909869 on Build GRCh38
rs529228518 Chr.1: 31909957-31909957 on Build GRCh38
rs892595364 Chr.1: 31909824-31909824 on Build GRCh38
rs947252262 Chr.1: 31909966-31909967 on Build GRCh38
rs182831250 Chr.1: 31910161-31910161 on Build GRCh38
rs572605655 Chr.1: 31909881-31909881 on Build GRCh38
rs148196214 Chr.1: 31910030-31910030 on Build GRCh38
rs753151051 Chr.1: 31910031-31910031 on Build GRCh38
rs745546362 Chr.1: 31910143-31910143 on Build GRCh38
rs756722451 Chr.1: 31910094-31910094 on Build GRCh38
rs917936260 Chr.1: 31909731-31909731 on Build GRCh38
rs776449108 Chr.1: 31910162-31910162 on Build GRCh38

View Full Product Details