Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 32266724-32267235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM1
Entrez Gene ID
64762
Gene Name
GRB2 associated regulator of MAPK1 subtype 1
Gene Aliases
C18orf11, FAM59A, GAREM, Gm944
UniGene
Hs.444314
Gene Chromosome Location
Chr.18: 32263521-32470518 on Build GRCh38
Transcripts
NM_001242409.1 NM_022751.2

View Full Product Details