Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 32267882-32268379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM1
Entrez Gene ID
64762
Gene Name
GRB2 associated regulator of MAPK1 subtype 1
Gene Aliases
C18orf11, FAM59A, GAREM, Gm944
UniGene
Hs.444314
Gene Chromosome Location
Chr.18: 32263521-32470518 on Build GRCh38
Transcripts
NM_001242409.1 NM_022751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5088523 c.2181delC p.R728fs*5 Chr.18: 32268321-32268321 on Build GRCh38
6293941 c.2335G>A p.A779T Chr.18: 32268167-32268167 on Build GRCh38
3525314 c.2356C>T p.P786S Chr.18: 32268146-32268146 on Build GRCh38
3525313 c.2559G>A p.E853E Chr.18: 32267943-32267943 on Build GRCh38
4478294 c.2228C>T p.P743L Chr.18: 32268274-32268274 on Build GRCh38
View More

View Full Product Details