Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 32268267-32268763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM1
Entrez Gene ID
64762
Gene Name
GRB2 associated regulator of MAPK1 subtype 1
Gene Aliases
C18orf11, FAM59A, GAREM, Gm944
UniGene
Hs.444314
Gene Chromosome Location
Chr.18: 32263521-32470518 on Build GRCh38
Transcripts
NM_001242409.1 NM_022751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756127 c.2029C>T p.L677L Chr.18: 32268473-32268473 on Build GRCh38
6669329 c.1814C>A p.S605Y Chr.18: 32268688-32268688 on Build GRCh38
6233469 c.2117C>A p.T706K Chr.18: 32268385-32268385 on Build GRCh38
5088523 c.2181delC p.R728fs*5 Chr.18: 32268321-32268321 on Build GRCh38
4473120 c.1829C>T p.S610F Chr.18: 32268673-32268673 on Build GRCh38
View More

View Full Product Details