Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 879407-879941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
UniGene
Hs.278973
Gene Chromosome Location
Chr.20: 869899-916367 on Build GRCh38
Transcripts
NM_001322809.1 NM_015985.3 XM_011529239.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1495118 c.1018G>A p.V340M Chr.20: 879782-879782 on Build GRCh38
5007679 c.985T>G p.W329G Chr.20: 879815-879815 on Build GRCh38
4401478 c.976G>A p.G326R Chr.20: 879824-879824 on Build GRCh38
5343664 c.1031G>A p.R344Q Chr.20: 879769-879769 on Build GRCh38
5392655 c.1041G>A p.K347K Chr.20: 879759-879759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150771715 Chr.20: 879753-879753 on Build GRCh38
rs775701631 Chr.20: 879797-879797 on Build GRCh38
rs778837477 Chr.20: 879815-879815 on Build GRCh38
rs557880338 Chr.20: 879770-879770 on Build GRCh38
rs61737018 Chr.20: 879783-879783 on Build GRCh38
rs529326916 Chr.20: 879509-879509 on Build GRCh38
rs745924075 Chr.20: 879461-879461 on Build GRCh38
rs139633349 Chr.20: 879765-879765 on Build GRCh38
rs1043238929 Chr.20: 879445-879445 on Build GRCh38
rs766803620 Chr.20: 879729-879729 on Build GRCh38
rs758523811 Chr.20: 879752-879752 on Build GRCh38
rs769951423 Chr.20: 879492-879492 on Build GRCh38
rs572371737 Chr.20: 879743-879743 on Build GRCh38
rs901370095 Chr.20: 879889-879889 on Build GRCh38
rs62641667 Chr.20: 879810-879810 on Build GRCh38
rs752366684 Chr.20: 879817-879817 on Build GRCh38
rs994295145 Chr.20: 879466-879466 on Build GRCh38
rs777242090 Chr.20: 879833-879833 on Build GRCh38
rs770189937 Chr.20: 879796-879796 on Build GRCh38
rs113886142 Chr.20: 879475-879475 on Build GRCh38
rs374341010 Chr.20: 879800-879800 on Build GRCh38
rs763322702 Chr.20: 879475-879475 on Build GRCh38
rs368408902 Chr.20: 879857-879857 on Build GRCh38
rs374881086 Chr.20: 879432-879432 on Build GRCh38
rs751529914 Chr.20: 879757-879757 on Build GRCh38
rs112915977 Chr.20: 879688-879688 on Build GRCh38
rs182945145 Chr.20: 879733-879733 on Build GRCh38
rs1021834185 Chr.20: 879508-879508 on Build GRCh38
rs1022198878 Chr.20: 879690-879690 on Build GRCh38
rs776992506 Chr.20: 879731-879731 on Build GRCh38
rs747724979 Chr.20: 879703-879703 on Build GRCh38
rs907239044 Chr.20: 879474-879474 on Build GRCh38
rs772585199 Chr.20: 879721-879721 on Build GRCh38
rs928728422 Chr.20: 879869-879869 on Build GRCh38
rs961255556 Chr.20: 879537-879537 on Build GRCh38
rs961791235 Chr.20: 879740-879740 on Build GRCh38
rs919796543 Chr.20: 879607-879607 on Build GRCh38
rs34557605 Chr.20: 879880-879880 on Build GRCh38
rs1047035601 Chr.20: 879471-879471 on Build GRCh38
rs1019882214 Chr.20: 879459-879459 on Build GRCh38
rs1042162080 Chr.20: 879638-879638 on Build GRCh38
rs113674912 Chr.20: 879667-879667 on Build GRCh38
rs746553557 Chr.20: 879835-879835 on Build GRCh38
rs146212103 Chr.20: 879578-879578 on Build GRCh38
rs377513353 Chr.20: 879767-879767 on Build GRCh38
rs760001649 Chr.20: 879789-879789 on Build GRCh38
rs770463987 Chr.20: 879852-879852 on Build GRCh38
rs756137911 Chr.20: 879762-879762 on Build GRCh38
rs559887554 Chr.20: 879622-879622 on Build GRCh38
rs771617269 Chr.20: 879778-879778 on Build GRCh38
rs55666732 Chr.20: 879508-879508 on Build GRCh38
rs903368694 Chr.20: 879698-879698 on Build GRCh38
rs781265643 Chr.20: 879582-879583 on Build GRCh38
rs774106865 Chr.20: 879701-879701 on Build GRCh38
rs763181713 Chr.20: 879900-879900 on Build GRCh38
rs1010700074 Chr.20: 879444-879444 on Build GRCh38
rs777341170 Chr.20: 879780-879780 on Build GRCh38
rs753000791 Chr.20: 879749-879749 on Build GRCh38
rs367908711 Chr.20: 879434-879434 on Build GRCh38
rs996843511 Chr.20: 879493-879493 on Build GRCh38
rs191056516 Chr.20: 879542-879542 on Build GRCh38
rs771662713 Chr.20: 879710-879710 on Build GRCh38
rs937950029 Chr.20: 879591-879591 on Build GRCh38
rs946415359 Chr.20: 879627-879627 on Build GRCh38
rs977859070 Chr.20: 879519-879519 on Build GRCh38
rs747895360 Chr.20: 879771-879771 on Build GRCh38
rs986680944 Chr.20: 879540-879540 on Build GRCh38
rs370994711 Chr.20: 879769-879769 on Build GRCh38
rs774574700 Chr.20: 879741-879741 on Build GRCh38
rs754954928 Chr.20: 879811-879811 on Build GRCh38
rs761949028 Chr.20: 879807-879807 on Build GRCh38
rs118083077 Chr.20: 879653-879653 on Build GRCh38
rs911028957 Chr.20: 879443-879443 on Build GRCh38
rs780654859 Chr.20: 879854-879854 on Build GRCh38
rs1329813 Chr.20: 879873-879873 on Build GRCh38
rs199734298 Chr.20: 879826-879826 on Build GRCh38
rs999473070 Chr.20: 879458-879458 on Build GRCh38
rs1054794560 Chr.20: 879599-879599 on Build GRCh38
rs758082232 Chr.20: 879829-879829 on Build GRCh38
rs567148755 Chr.20: 879665-879665 on Build GRCh38
rs952720465 Chr.20: 879723-879723 on Build GRCh38
rs555627321 Chr.20: 879699-879699 on Build GRCh38
rs759844391 Chr.20: 879732-879732 on Build GRCh38
rs566006653 Chr.20: 879559-879559 on Build GRCh38
rs543898521 Chr.20: 879507-879507 on Build GRCh38
rs1028302161 Chr.20: 879494-879494 on Build GRCh38
rs111505550 Chr.20: 879649-879649 on Build GRCh38
rs773747151 Chr.20: 879870-879870 on Build GRCh38
rs1012162351 Chr.20: 879507-879508 on Build GRCh38
rs147649307 Chr.20: 879799-879799 on Build GRCh38
rs746429080 Chr.20: 879855-879855 on Build GRCh38
rs746445808 Chr.20: 879782-879782 on Build GRCh38
rs903359850 Chr.20: 879446-879446 on Build GRCh38
rs528567403 Chr.20: 879654-879654 on Build GRCh38
rs374903754 Chr.20: 879893-879893 on Build GRCh38
rs141810723 Chr.20: 879538-879539 on Build GRCh38
rs761310089 Chr.20: 879724-879724 on Build GRCh38

View Full Product Details