Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 915828-916336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
UniGene
Hs.278973
Gene Chromosome Location
Chr.20: 869899-916367 on Build GRCh38
Transcripts
NM_001322809.1 NM_015985.3 XM_011529239.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3379129 c.308A>G p.K103R Chr.20: 915907-915907 on Build GRCh38
3841599 c.260T>G p.V87G Chr.20: 915955-915955 on Build GRCh38
478419 c.225C>T p.N75N Chr.20: 915990-915990 on Build GRCh38
3841600 c.227C>A p.P76Q Chr.20: 915988-915988 on Build GRCh38
478420 c.148C>T p.P50S Chr.20: 916067-916067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892630652 Chr.20: 916000-916000 on Build GRCh38
rs757405231 Chr.20: 915963-915963 on Build GRCh38
rs376690664 Chr.20: 916260-916260 on Build GRCh38
rs925274892 Chr.20: 915925-915925 on Build GRCh38
rs777726255 Chr.20: 915888-915888 on Build GRCh38
rs140900541 Chr.20: 916015-916015 on Build GRCh38
rs199745156 Chr.20: 915936-915936 on Build GRCh38
rs199886060 Chr.20: 916099-916099 on Build GRCh38
rs562175835 Chr.20: 915853-915853 on Build GRCh38
rs369053720 Chr.20: 916129-916129 on Build GRCh38
rs749934902 Chr.20: 915921-915921 on Build GRCh38
rs774515703 Chr.20: 916042-916055 on Build GRCh38
rs201487067 Chr.20: 916223-916223 on Build GRCh38
rs752749565 Chr.20: 916162-916162 on Build GRCh38
rs761386387 Chr.20: 915917-915917 on Build GRCh38
rs768523640 Chr.20: 915978-915978 on Build GRCh38
rs377149306 Chr.20: 915922-915922 on Build GRCh38
rs768424182 Chr.20: 915902-915903 on Build GRCh38
rs200170732 Chr.20: 916147-916147 on Build GRCh38
rs945525695 Chr.20: 916315-916315 on Build GRCh38
rs144442482 Chr.20: 916045-916045 on Build GRCh38
rs574115140 Chr.20: 915990-915990 on Build GRCh38
rs753048955 Chr.20: 915948-915948 on Build GRCh38
rs139001653 Chr.20: 915947-915947 on Build GRCh38
rs746681989 Chr.20: 916095-916095 on Build GRCh38
rs73565266 Chr.20: 915902-915902 on Build GRCh38
rs960876006 Chr.20: 915890-915890 on Build GRCh38
rs764806114 Chr.20: 916116-916116 on Build GRCh38
rs772655243 Chr.20: 916252-916253 on Build GRCh38
rs369518796 Chr.20: 915870-915870 on Build GRCh38
rs146594844 Chr.20: 916072-916072 on Build GRCh38
rs372508631 Chr.20: 916178-916178 on Build GRCh38
rs750615088 Chr.20: 916134-916134 on Build GRCh38
rs370905563 Chr.20: 916233-916233 on Build GRCh38
rs905433662 Chr.20: 915966-915966 on Build GRCh38
rs972144749 Chr.20: 916218-916219 on Build GRCh38
rs748015813 Chr.20: 916084-916084 on Build GRCh38
rs771735517 Chr.20: 915991-915991 on Build GRCh38
rs771963050 Chr.20: 916087-916087 on Build GRCh38
rs146463028 Chr.20: 915998-915998 on Build GRCh38
rs758206666 Chr.20: 916184-916184 on Build GRCh38
rs776112183 Chr.20: 916118-916118 on Build GRCh38
rs762854677 Chr.20: 915901-915901 on Build GRCh38
rs376436931 Chr.20: 916229-916229 on Build GRCh38
rs201720367 Chr.20: 916029-916029 on Build GRCh38
rs763353908 Chr.20: 916122-916122 on Build GRCh38
rs774067368 Chr.20: 915915-915915 on Build GRCh38
rs770735432 Chr.20: 916221-916221 on Build GRCh38
rs772883230 Chr.20: 916094-916094 on Build GRCh38
rs764585947 Chr.20: 916128-916128 on Build GRCh38
rs749289515 Chr.20: 915968-915968 on Build GRCh38
rs969626509 Chr.20: 916160-916160 on Build GRCh38
rs933436193 Chr.20: 916254-916254 on Build GRCh38
rs745899909 Chr.20: 916050-916050 on Build GRCh38
rs762278397 Chr.20: 916048-916048 on Build GRCh38
rs758569315 Chr.20: 915955-915955 on Build GRCh38
rs747058723 Chr.20: 915897-915897 on Build GRCh38
rs781235665 Chr.20: 915966-915966 on Build GRCh38
rs778013651 Chr.20: 915959-915959 on Build GRCh38
rs764341218 Chr.20: 916032-916032 on Build GRCh38
rs1020061490 Chr.20: 916026-916026 on Build GRCh38
rs774153532 Chr.20: 915986-915986 on Build GRCh38
rs769649352 Chr.20: 915898-915898 on Build GRCh38
rs776299477 Chr.20: 916106-916106 on Build GRCh38
rs746729787 Chr.20: 916196-916196 on Build GRCh38
rs753031251 Chr.20: 916025-916025 on Build GRCh38
rs369649721 Chr.20: 915896-915896 on Build GRCh38
rs755540821 Chr.20: 915944-915944 on Build GRCh38
rs777640954 Chr.20: 916191-916191 on Build GRCh38
rs936958143 Chr.20: 915994-915994 on Build GRCh38
rs757415116 Chr.20: 916044-916044 on Build GRCh38
rs776217811 Chr.20: 916010-916010 on Build GRCh38
rs865857735 Chr.20: 915918-915918 on Build GRCh38
rs778863210 Chr.20: 916083-916083 on Build GRCh38
rs772265766 Chr.20: 915879-915879 on Build GRCh38
rs772969383 Chr.20: 915993-915993 on Build GRCh38
rs754369241 Chr.20: 915869-915869 on Build GRCh38
rs200815484 Chr.20: 916043-916043 on Build GRCh38
rs1037597430 Chr.20: 916297-916297 on Build GRCh38
rs770645507 Chr.20: 916101-916101 on Build GRCh38
rs756603067 Chr.20: 915858-915858 on Build GRCh38
rs993180137 Chr.20: 916089-916089 on Build GRCh38
rs373183124 Chr.20: 916234-916234 on Build GRCh38
rs774809629 Chr.20: 916248-916248 on Build GRCh38
rs980098439 Chr.20: 916170-916170 on Build GRCh38
rs746957688 Chr.20: 916082-916082 on Build GRCh38
rs765788530 Chr.20: 916019-916019 on Build GRCh38
rs762045720 Chr.20: 916130-916130 on Build GRCh38
rs370336882 Chr.20: 915861-915861 on Build GRCh38
rs914045752 Chr.20: 916310-916310 on Build GRCh38
rs775445001 Chr.20: 915900-915900 on Build GRCh38
rs17702306 Chr.20: 916110-916110 on Build GRCh38
rs933148834 Chr.20: 915965-915965 on Build GRCh38
rs116327782 Chr.20: 915931-915931 on Build GRCh38
rs773882603 Chr.20: 916265-916278 on Build GRCh38
rs767686759 Chr.20: 916131-916131 on Build GRCh38
rs149874461 Chr.20: 915962-915962 on Build GRCh38
rs953311251 Chr.20: 916074-916074 on Build GRCh38
rs760234079 Chr.20: 915995-915995 on Build GRCh38
rs748445216 Chr.20: 915871-915871 on Build GRCh38
rs756237515 Chr.20: 916145-916145 on Build GRCh38
rs779387259 Chr.20: 916275-916275 on Build GRCh38
rs374092321 Chr.20: 916264-916264 on Build GRCh38

View Full Product Details