Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159993442-159993968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
UniGene
Hs.673
Gene Chromosome Location
Chr.3: 159988836-159996019 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Gene Symbol
IL12A-AS1
Entrez Gene ID
101928376
Gene Name
IL12A antisense RNA 1
Gene Aliases
-
UniGene
Hs.565865
Gene Chromosome Location
Chr.3: 159913400-160207092 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4832004 c.410C>G p.S137C Chr.3: 159993482-159993482 on Build GRCh38
3589426 c.554G>A p.R185K Chr.3: 159993792-159993792 on Build GRCh38
1617136 c.559A>G p.I187V Chr.3: 159993797-159993797 on Build GRCh38
6684831 c.387T>C p.S129S Chr.3: 159993459-159993459 on Build GRCh38
7200036 c.384G>T p.E128D Chr.3: 159993456-159993456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149846802 Chr.3: 159993600-159993600 on Build GRCh38
rs773136641 Chr.3: 159993806-159993806 on Build GRCh38
rs751284391 Chr.3: 159993519-159993520 on Build GRCh38
rs761130017 Chr.3: 159993564-159993564 on Build GRCh38
rs756092828 Chr.3: 159993640-159993640 on Build GRCh38
rs923023325 Chr.3: 159993559-159993559 on Build GRCh38
rs765267336 Chr.3: 159993875-159993875 on Build GRCh38
rs765492561 Chr.3: 159993592-159993592 on Build GRCh38
rs748559121 Chr.3: 159993791-159993791 on Build GRCh38
rs765173014 Chr.3: 159993690-159993690 on Build GRCh38
rs913028578 Chr.3: 159993505-159993505 on Build GRCh38
rs76693142 Chr.3: 159993921-159993921 on Build GRCh38
rs1024513737 Chr.3: 159993815-159993815 on Build GRCh38
rs538084986 Chr.3: 159993843-159993843 on Build GRCh38
rs781551474 Chr.3: 159993764-159993764 on Build GRCh38
rs763929222 Chr.3: 159993852-159993852 on Build GRCh38
rs779055728 Chr.3: 159993470-159993470 on Build GRCh38
rs990104054 Chr.3: 159993497-159993497 on Build GRCh38
rs748799140 Chr.3: 159993647-159993647 on Build GRCh38
rs190538535 Chr.3: 159993661-159993661 on Build GRCh38
rs754797956 Chr.3: 159993487-159993487 on Build GRCh38
rs574173919 Chr.3: 159993810-159993810 on Build GRCh38
rs771787311 Chr.3: 159993655-159993655 on Build GRCh38
rs976811495 Chr.3: 159993861-159993861 on Build GRCh38
rs756428573 Chr.3: 159993526-159993527 on Build GRCh38
rs779545994 Chr.3: 159993885-159993885 on Build GRCh38
rs773368346 Chr.3: 159993691-159993691 on Build GRCh38
rs754648444 Chr.3: 159993895-159993895 on Build GRCh38
rs762313783 Chr.3: 159993684-159993684 on Build GRCh38
rs750544982 Chr.3: 159993594-159993594 on Build GRCh38
rs201655135 Chr.3: 159993478-159993478 on Build GRCh38
rs565337869 Chr.3: 159993703-159993703 on Build GRCh38
rs375527576 Chr.3: 159993509-159993509 on Build GRCh38
rs749886054 Chr.3: 159993882-159993882 on Build GRCh38
rs756037063 Chr.3: 159993704-159993704 on Build GRCh38
rs776668796 Chr.3: 159993582-159993582 on Build GRCh38
rs866340019 Chr.3: 159993475-159993475 on Build GRCh38
rs773148606 Chr.3: 159993823-159993823 on Build GRCh38
rs535488207 Chr.3: 159993905-159993906 on Build GRCh38
rs779949936 Chr.3: 159993518-159993519 on Build GRCh38
rs745535302 Chr.3: 159993547-159993547 on Build GRCh38
rs775946182 Chr.3: 159993557-159993557 on Build GRCh38
rs764623851 Chr.3: 159993573-159993573 on Build GRCh38
rs2230052 Chr.3: 159993805-159993805 on Build GRCh38
rs369508050 Chr.3: 159993870-159993870 on Build GRCh38
rs763048900 Chr.3: 159993694-159993694 on Build GRCh38
rs935618466 Chr.3: 159993880-159993880 on Build GRCh38
rs778132771 Chr.3: 159993532-159993534 on Build GRCh38
rs9832255 Chr.3: 159993541-159993541 on Build GRCh38
rs748044888 Chr.3: 159993507-159993507 on Build GRCh38
rs777190597 Chr.3: 159993679-159993679 on Build GRCh38
rs988377448 Chr.3: 159993903-159993903 on Build GRCh38
rs749466612 Chr.3: 159993626-159993626 on Build GRCh38
rs1040208561 Chr.3: 159993549-159993549 on Build GRCh38
rs545507796 Chr.3: 159993905-159993905 on Build GRCh38
rs766667606 Chr.3: 159993701-159993701 on Build GRCh38
rs772263185 Chr.3: 159993552-159993552 on Build GRCh38
rs778246375 Chr.3: 159993737-159993737 on Build GRCh38
rs778494399 Chr.3: 159993653-159993653 on Build GRCh38
rs556606966 Chr.3: 159993844-159993844 on Build GRCh38
rs756651610 Chr.3: 159993652-159993652 on Build GRCh38
rs375505799 Chr.3: 159993519-159993519 on Build GRCh38
rs749989170 Chr.3: 159993745-159993745 on Build GRCh38
rs773938409 Chr.3: 159993536-159993536 on Build GRCh38
rs964398253 Chr.3: 159993833-159993833 on Build GRCh38
rs770692194 Chr.3: 159993525-159993525 on Build GRCh38
rs1001624071 Chr.3: 159993827-159993827 on Build GRCh38
rs186843156 Chr.3: 159993606-159993606 on Build GRCh38
rs753767038 Chr.3: 159993723-159993723 on Build GRCh38
rs758664036 Chr.3: 159993599-159993599 on Build GRCh38
rs538740981 Chr.3: 159993643-159993643 on Build GRCh38
rs753716045 Chr.3: 159993860-159993860 on Build GRCh38
rs748850178 Chr.3: 159993522-159993522 on Build GRCh38
rs768944534 Chr.3: 159993760-159993760 on Build GRCh38
rs201524139 Chr.3: 159993883-159993883 on Build GRCh38
rs747421632 Chr.3: 159993668-159993668 on Build GRCh38
rs769140786 Chr.3: 159993678-159993678 on Build GRCh38
rs757963512 Chr.3: 159993748-159993748 on Build GRCh38
rs757237511 Chr.3: 159993724-159993724 on Build GRCh38
rs55780930 Chr.3: 159993786-159993786 on Build GRCh38
rs763982444 Chr.3: 159993711-159993711 on Build GRCh38
rs761941291 Chr.3: 159993586-159993586 on Build GRCh38
rs752574623 Chr.3: 159993618-159993618 on Build GRCh38
rs751154856 Chr.3: 159993894-159993894 on Build GRCh38
rs757414291 Chr.3: 159993678-159993678 on Build GRCh38
rs534180973 Chr.3: 159993749-159993749 on Build GRCh38
rs374624685 Chr.3: 159993501-159993501 on Build GRCh38

View Full Product Details