Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159995656-159996159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
UniGene
Hs.673
Gene Chromosome Location
Chr.3: 159988836-159996019 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Gene Symbol
IL12A-AS1
Entrez Gene ID
101928376
Gene Name
IL12A antisense RNA 1
Gene Aliases
-
UniGene
Hs.565865
Gene Chromosome Location
Chr.3: 159913400-160207092 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

SNP Detail

SNP ID Chromosome Location
rs1014265099 Chr.3: 159995996-159995996 on Build GRCh38
rs1021994260 Chr.3: 159996083-159996083 on Build GRCh38
rs11421436 Chr.3: 159995729-159995730 on Build GRCh38
rs982512290 Chr.3: 159995729-159995729 on Build GRCh38
rs566278676 Chr.3: 159995838-159995838 on Build GRCh38
rs933932946 Chr.3: 159995796-159995796 on Build GRCh38
rs969037447 Chr.3: 159995716-159995716 on Build GRCh38
rs899072345 Chr.3: 159995935-159995935 on Build GRCh38
rs929249525 Chr.3: 159995946-159995946 on Build GRCh38
rs777319942 Chr.3: 159995764-159995765 on Build GRCh38
rs1037749183 Chr.3: 159995896-159995896 on Build GRCh38
rs748797323 Chr.3: 159995923-159995924 on Build GRCh38
rs568408 Chr.3: 159995680-159995680 on Build GRCh38
rs1016385843 Chr.3: 159996081-159996082 on Build GRCh38
rs924789518 Chr.3: 159995791-159995791 on Build GRCh38
rs1026467429 Chr.3: 159995688-159995688 on Build GRCh38
rs536702657 Chr.3: 159996024-159996024 on Build GRCh38
rs6774042 Chr.3: 159996048-159996048 on Build GRCh38
rs977550602 Chr.3: 159995757-159995757 on Build GRCh38
rs1003263596 Chr.3: 159996030-159996030 on Build GRCh38
rs372535187 Chr.3: 159996017-159996017 on Build GRCh38
rs938106513 Chr.3: 159995952-159995955 on Build GRCh38
rs943346990 Chr.3: 159995853-159995853 on Build GRCh38
rs640039 Chr.3: 159996059-159996059 on Build GRCh38
rs903968555 Chr.3: 159995940-159995940 on Build GRCh38
rs986687624 Chr.3: 159995800-159995800 on Build GRCh38
rs547979553 Chr.3: 159995837-159995837 on Build GRCh38
rs975524835 Chr.3: 159995763-159995763 on Build GRCh38
rs896470792 Chr.3: 159995972-159995972 on Build GRCh38
rs1047557880 Chr.3: 159995965-159995965 on Build GRCh38
rs1016598427 Chr.3: 159995735-159995735 on Build GRCh38
rs866927196 Chr.3: 159995753-159995753 on Build GRCh38
rs933384699 Chr.3: 159995826-159995826 on Build GRCh38
rs920924312 Chr.3: 159995764-159995764 on Build GRCh38
rs1022274172 Chr.3: 159995692-159995692 on Build GRCh38
rs910666206 Chr.3: 159995845-159995845 on Build GRCh38
rs116287698 Chr.3: 159996038-159996038 on Build GRCh38

View Full Product Details