Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 176711307-176711832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VEGFC
Entrez Gene ID
7424
Gene Name
vascular endothelial growth factor C
Gene Aliases
Flt4-L, LMPH1D, VRP
UniGene
Hs.435215
Gene Chromosome Location
Chr.4: 176683534-176792745 on Build GRCh38
Transcripts
NM_005429.4

SNP Detail

SNP ID Chromosome Location
rs766629934 Chr.4: 176711678-176711678 on Build GRCh38
rs762630317 Chr.4: 176711510-176711510 on Build GRCh38
rs769325515 Chr.4: 176711500-176711500 on Build GRCh38
rs924316826 Chr.4: 176711657-176711657 on Build GRCh38
rs565818581 Chr.4: 176711717-176711717 on Build GRCh38
rs748640295 Chr.4: 176711593-176711593 on Build GRCh38
rs547569778 Chr.4: 176711424-176711424 on Build GRCh38
rs758469771 Chr.4: 176711675-176711675 on Build GRCh38
rs781700431 Chr.4: 176711581-176711581 on Build GRCh38
rs962853807 Chr.4: 176711793-176711793 on Build GRCh38
rs748908979 Chr.4: 176711630-176711630 on Build GRCh38
rs763741167 Chr.4: 176711353-176711353 on Build GRCh38
rs755358775 Chr.4: 176711681-176711681 on Build GRCh38
rs779214732 Chr.4: 176711668-176711668 on Build GRCh38
rs574723843 Chr.4: 176711797-176711797 on Build GRCh38
rs534942500 Chr.4: 176711738-176711738 on Build GRCh38
rs752645297 Chr.4: 176711541-176711541 on Build GRCh38
rs1019209542 Chr.4: 176711350-176711350 on Build GRCh38
rs796701107 Chr.4: 176711604-176711604 on Build GRCh38
rs759440089 Chr.4: 176711647-176711647 on Build GRCh38
rs186533755 Chr.4: 176711551-176711551 on Build GRCh38
rs970187597 Chr.4: 176711602-176711602 on Build GRCh38
rs751935960 Chr.4: 176711420-176711423 on Build GRCh38
rs955780450 Chr.4: 176711371-176711371 on Build GRCh38
rs747518311 Chr.4: 176711457-176711457 on Build GRCh38
rs769210473 Chr.4: 176711459-176711459 on Build GRCh38
rs538678541 Chr.4: 176711598-176711598 on Build GRCh38
rs777120974 Chr.4: 176711471-176711471 on Build GRCh38
rs867871455 Chr.4: 176711662-176711662 on Build GRCh38
rs35739654 Chr.4: 176711698-176711699 on Build GRCh38
rs377096543 Chr.4: 176711574-176711574 on Build GRCh38
rs775311204 Chr.4: 176711658-176711658 on Build GRCh38
rs527847482 Chr.4: 176711360-176711360 on Build GRCh38
rs993778924 Chr.4: 176711724-176711724 on Build GRCh38
rs181841068 Chr.4: 176711474-176711474 on Build GRCh38
rs368621055 Chr.4: 176711700-176711700 on Build GRCh38
rs766283110 Chr.4: 176711512-176711512 on Build GRCh38
rs554798707 Chr.4: 176711796-176711796 on Build GRCh38
rs991354078 Chr.4: 176711777-176711777 on Build GRCh38
rs760621275 Chr.4: 176711659-176711659 on Build GRCh38
rs774139549 Chr.4: 176711632-176711632 on Build GRCh38
rs780173266 Chr.4: 176711754-176711754 on Build GRCh38
rs200933230 Chr.4: 176711544-176711544 on Build GRCh38
rs772765057 Chr.4: 176711506-176711506 on Build GRCh38
rs753912137 Chr.4: 176711675-176711675 on Build GRCh38
rs192007157 Chr.4: 176711674-176711674 on Build GRCh38
rs983059268 Chr.4: 176711404-176711404 on Build GRCh38
rs1017173042 Chr.4: 176711467-176711467 on Build GRCh38
rs1024114016 Chr.4: 176711778-176711778 on Build GRCh38
rs587777567 Chr.4: 176711575-176711575 on Build GRCh38
rs374006758 Chr.4: 176711569-176711569 on Build GRCh38
rs370267488 Chr.4: 176711665-176711665 on Build GRCh38
rs750642281 Chr.4: 176711549-176711549 on Build GRCh38
rs565462956 Chr.4: 176711348-176711348 on Build GRCh38
rs981820310 Chr.4: 176711608-176711608 on Build GRCh38
rs45555032 Chr.4: 176711355-176711355 on Build GRCh38
rs550015862 Chr.4: 176711520-176711520 on Build GRCh38
rs1038739455 Chr.4: 176711684-176711684 on Build GRCh38
rs767394771 Chr.4: 176711648-176711648 on Build GRCh38
rs774233934 Chr.4: 176711515-176711515 on Build GRCh38
rs760511238 Chr.4: 176711347-176711347 on Build GRCh38
rs760870718 Chr.4: 176711473-176711473 on Build GRCh38
rs751758352 Chr.4: 176711680-176711680 on Build GRCh38
rs1026311065 Chr.4: 176711725-176711725 on Build GRCh38
rs772306595 Chr.4: 176711750-176711750 on Build GRCh38
rs770761859 Chr.4: 176711631-176711631 on Build GRCh38
rs561174157 Chr.4: 176711461-176711461 on Build GRCh38
rs949696109 Chr.4: 176711613-176711613 on Build GRCh38
rs997285004 Chr.4: 176711726-176711726 on Build GRCh38
rs952090483 Chr.4: 176711400-176711400 on Build GRCh38
rs767627714 Chr.4: 176711533-176711533 on Build GRCh38
rs750457371 Chr.4: 176711673-176711673 on Build GRCh38
rs1015882935 Chr.4: 176711783-176711783 on Build GRCh38
rs928660549 Chr.4: 176711645-176711645 on Build GRCh38
rs368039767 Chr.4: 176711695-176711695 on Build GRCh38
rs765310637 Chr.4: 176711548-176711548 on Build GRCh38
rs772716675 Chr.4: 176711617-176711617 on Build GRCh38
rs764201347 Chr.4: 176711472-176711473 on Build GRCh38
rs1053005535 Chr.4: 176711663-176711663 on Build GRCh38
rs371968637 Chr.4: 176711799-176711799 on Build GRCh38
rs587777566 Chr.4: 176711631-176711632 on Build GRCh38
rs780409353 Chr.4: 176711561-176711561 on Build GRCh38

View Full Product Details