Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243501195-243501692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT3
Entrez Gene ID
10000
Gene Name
AKT serine/threonine kinase 3
Gene Aliases
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2
UniGene
Hs.498292
Gene Chromosome Location
Chr.1: 243488233-243851079 on Build GRCh38
Transcripts
NM_001206729.1 NM_005465.4 NM_181690.2 XM_005272994.4 XM_005272995.2

SNP Detail

SNP ID Chromosome Location
rs75847692 Chr.1: 243501518-243501518 on Build GRCh38
rs138661915 Chr.1: 243501244-243501244 on Build GRCh38
rs16356 Chr.1: 243501623-243501624 on Build GRCh38
rs933500303 Chr.1: 243501254-243501254 on Build GRCh38
rs192437955 Chr.1: 243501559-243501559 on Build GRCh38
rs113544916 Chr.1: 243501312-243501312 on Build GRCh38
rs537527172 Chr.1: 243501576-243501576 on Build GRCh38
rs756172746 Chr.1: 243501411-243501411 on Build GRCh38
rs925558493 Chr.1: 243501235-243501235 on Build GRCh38
rs547675480 Chr.1: 243501432-243501432 on Build GRCh38
rs933511599 Chr.1: 243501450-243501450 on Build GRCh38
rs979174570 Chr.1: 243501311-243501311 on Build GRCh38
rs937058305 Chr.1: 243501633-243501633 on Build GRCh38
rs780337464 Chr.1: 243501545-243501545 on Build GRCh38
rs974710553 Chr.1: 243501354-243501354 on Build GRCh38
rs767170769 Chr.1: 243501414-243501415 on Build GRCh38
rs1023689924 Chr.1: 243501365-243501365 on Build GRCh38
rs149318674 Chr.1: 243501261-243501261 on Build GRCh38
rs979406368 Chr.1: 243501370-243501370 on Build GRCh38
rs1058305 Chr.1: 243501340-243501340 on Build GRCh38
rs372382366 Chr.1: 243501599-243501600 on Build GRCh38
rs368033370 Chr.1: 243501621-243501621 on Build GRCh38
rs535815351 Chr.1: 243501441-243501441 on Build GRCh38
rs1002177974 Chr.1: 243501478-243501478 on Build GRCh38
rs763016492 Chr.1: 243501311-243501312 on Build GRCh38
rs898195788 Chr.1: 243501537-243501537 on Build GRCh38
rs923465631 Chr.1: 243501253-243501253 on Build GRCh38
rs184373409 Chr.1: 243501510-243501510 on Build GRCh38
rs778161726 Chr.1: 243501669-243501669 on Build GRCh38
rs568378973 Chr.1: 243501234-243501234 on Build GRCh38
rs188241383 Chr.1: 243501369-243501369 on Build GRCh38
rs192979757 Chr.1: 243501424-243501424 on Build GRCh38
rs1032383625 Chr.1: 243501623-243501623 on Build GRCh38
rs974621882 Chr.1: 243501560-243501560 on Build GRCh38
rs750973612 Chr.1: 243501308-243501308 on Build GRCh38
rs896334913 Chr.1: 243501362-243501362 on Build GRCh38
rs561325233 Chr.1: 243501598-243501598 on Build GRCh38
rs988332662 Chr.1: 243501245-243501245 on Build GRCh38
rs534852128 Chr.1: 243501300-243501300 on Build GRCh38
rs1020416150 Chr.1: 243501306-243501306 on Build GRCh38
rs905296382 Chr.1: 243501599-243501599 on Build GRCh38
rs373176480 Chr.1: 243501422-243501422 on Build GRCh38
rs1037481653 Chr.1: 243501616-243501616 on Build GRCh38
rs572502589 Chr.1: 243501459-243501459 on Build GRCh38
rs550016430 Chr.1: 243501283-243501283 on Build GRCh38
rs994526099 Chr.1: 243501314-243501314 on Build GRCh38
rs556335908 Chr.1: 243501401-243501401 on Build GRCh38
rs541396136 Chr.1: 243501596-243501596 on Build GRCh38
rs531561178 Chr.1: 243501260-243501260 on Build GRCh38
rs548162696 Chr.1: 243501238-243501239 on Build GRCh38
rs907631466 Chr.1: 243501431-243501431 on Build GRCh38
rs527315754 Chr.1: 243501664-243501664 on Build GRCh38
rs986269094 Chr.1: 243501468-243501468 on Build GRCh38
rs189486611 Chr.1: 243501511-243501511 on Build GRCh38
rs1058304 Chr.1: 243501555-243501555 on Build GRCh38
rs370563834 Chr.1: 243501448-243501448 on Build GRCh38
rs766074349 Chr.1: 243501288-243501288 on Build GRCh38
rs547790258 Chr.1: 243501219-243501219 on Build GRCh38
rs1013408640 Chr.1: 243501364-243501364 on Build GRCh38
rs933188304 Chr.1: 243501399-243501399 on Build GRCh38
rs895981445 Chr.1: 243501493-243501493 on Build GRCh38
rs397831754 Chr.1: 243501629-243501630 on Build GRCh38
rs987271306 Chr.1: 243501597-243501598 on Build GRCh38
rs1056089607 Chr.1: 243501480-243501480 on Build GRCh38
rs907598981 Chr.1: 243501598-243501599 on Build GRCh38

View Full Product Details