Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243502774-243503267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT3
Entrez Gene ID
10000
Gene Name
AKT serine/threonine kinase 3
Gene Aliases
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2
UniGene
Hs.498292
Gene Chromosome Location
Chr.1: 243488233-243851079 on Build GRCh38
Transcripts
NM_001206729.1 NM_005465.4 NM_181690.2 XM_005272994.4 XM_005272995.2 XM_011544014.2 XM_016999985.1

SNP Detail

SNP ID Chromosome Location
rs954174200 Chr.1: 243503130-243503130 on Build GRCh38
rs957290949 Chr.1: 243503122-243503122 on Build GRCh38
rs911983788 Chr.1: 243503240-243503240 on Build GRCh38
rs564075522 Chr.1: 243503179-243503179 on Build GRCh38
rs181761083 Chr.1: 243502968-243502968 on Build GRCh38
rs185750644 Chr.1: 243503102-243503102 on Build GRCh38
rs755232002 Chr.1: 243502915-243502915 on Build GRCh38
rs897712672 Chr.1: 243502930-243502930 on Build GRCh38
rs957660055 Chr.1: 243502806-243502806 on Build GRCh38
rs372540763 Chr.1: 243503052-243503052 on Build GRCh38
rs1037049717 Chr.1: 243502929-243502929 on Build GRCh38
rs990829378 Chr.1: 243502845-243502845 on Build GRCh38
rs926558903 Chr.1: 243502852-243502852 on Build GRCh38
rs781207359 Chr.1: 243502963-243502963 on Build GRCh38
rs774370271 Chr.1: 243502900-243502902 on Build GRCh38
rs570624987 Chr.1: 243503138-243503138 on Build GRCh38
rs1014821867 Chr.1: 243503048-243503048 on Build GRCh38
rs563859604 Chr.1: 243502939-243502939 on Build GRCh38
rs531329460 Chr.1: 243503217-243503217 on Build GRCh38
rs199913818 Chr.1: 243502895-243502896 on Build GRCh38
rs748250019 Chr.1: 243503003-243503003 on Build GRCh38
rs1051885780 Chr.1: 243502945-243502945 on Build GRCh38
rs1033272588 Chr.1: 243503118-243503118 on Build GRCh38
rs995135617 Chr.1: 243502991-243502991 on Build GRCh38
rs145577971 Chr.1: 243502979-243502979 on Build GRCh38
rs1024026532 Chr.1: 243503050-243503050 on Build GRCh38
rs7521027 Chr.1: 243503034-243503034 on Build GRCh38
rs879188187 Chr.1: 243503101-243503102 on Build GRCh38
rs189812314 Chr.1: 243502928-243502928 on Build GRCh38
rs769595591 Chr.1: 243503177-243503177 on Build GRCh38
rs932858106 Chr.1: 243502889-243502890 on Build GRCh38
rs555005002 Chr.1: 243503205-243503205 on Build GRCh38
rs1049484119 Chr.1: 243502956-243502956 on Build GRCh38
rs751781333 Chr.1: 243502896-243502896 on Build GRCh38
rs1024690912 Chr.1: 243503063-243503063 on Build GRCh38
rs936551997 Chr.1: 243502864-243502864 on Build GRCh38
rs539325565 Chr.1: 243503154-243503154 on Build GRCh38
rs190950461 Chr.1: 243503186-243503186 on Build GRCh38
rs957934327 Chr.1: 243503126-243503126 on Build GRCh38
rs943370382 Chr.1: 243502951-243502951 on Build GRCh38
rs931345244 Chr.1: 243503218-243503218 on Build GRCh38
rs940139371 Chr.1: 243503163-243503163 on Build GRCh38
rs535312241 Chr.1: 243503202-243503202 on Build GRCh38
rs890516916 Chr.1: 243502946-243502946 on Build GRCh38
rs970426157 Chr.1: 243503092-243503092 on Build GRCh38
rs760538042 Chr.1: 243503059-243503061 on Build GRCh38
rs551766297 Chr.1: 243503103-243503103 on Build GRCh38
rs924827846 Chr.1: 243502795-243502795 on Build GRCh38
rs1026982631 Chr.1: 243503031-243503031 on Build GRCh38
rs912618614 Chr.1: 243503145-243503145 on Build GRCh38
rs550525204 Chr.1: 243502793-243502793 on Build GRCh38
rs552114137 Chr.1: 243503126-243503129 on Build GRCh38
rs989726462 Chr.1: 243502884-243502884 on Build GRCh38
rs1048967642 Chr.1: 243503228-243503228 on Build GRCh38

View Full Product Details