Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 533666-534164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRAS
Entrez Gene ID
3265
Gene Name
HRas proto-oncogene, GTPase
Gene Aliases
C-BAS/HAS, C-H-RAS, C-HA-RAS1, CTLO, H-RASIDX, HAMSV, HRAS1, RASH1, p21ras
UniGene
Hs.37003
Gene Chromosome Location
Chr.11: 532242-535567 on Build GRCh38
Transcripts
NM_001130442.2 NM_001318054.1 NM_005343.3 NM_176795.4 XM_011519875.2 XM_011519877.2 XM_017017167.1 XM_017017168.1

Gene Symbol
LRRC56
Entrez Gene ID
115399
Gene Name
leucine rich repeat containing 56
Gene Aliases
-
UniGene
Hs.567655
Gene Chromosome Location
Chr.11: 518967-554916 on Build GRCh38
Transcripts
NM_001130442.2 NM_001318054.1 NM_005343.3 NM_176795.4 XM_011519875.2 XM_011519877.2 XM_017017167.1 XM_017017168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6854315 c.190T>C p.Y64H Chr.11: 533866-533866 on Build GRCh38
6929976 c.203G>A p.R68Q Chr.11: 533853-533853 on Build GRCh38
4471782 c.173C>T p.T58I Chr.11: 533883-533883 on Build GRCh38
3687359 c.222C>T p.T74T Chr.11: 533834-533834 on Build GRCh38
6937842 c.218G>A p.R73H Chr.11: 533838-533838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560393669 Chr.11: 533735-533735 on Build GRCh38
rs530529842 Chr.11: 533706-533706 on Build GRCh38
rs992133771 Chr.11: 534006-534006 on Build GRCh38
rs370318085 Chr.11: 533715-533715 on Build GRCh38
rs755322824 Chr.11: 533790-533790 on Build GRCh38
rs746697013 Chr.11: 533981-533981 on Build GRCh38
rs751844306 Chr.11: 533733-533733 on Build GRCh38
rs765092617 Chr.11: 533915-533915 on Build GRCh38
rs896550504 Chr.11: 534113-534113 on Build GRCh38
rs923755318 Chr.11: 534051-534051 on Build GRCh38
rs879158955 Chr.11: 534075-534075 on Build GRCh38
rs755577443 Chr.11: 533963-533963 on Build GRCh38
rs771774973 Chr.11: 533857-533857 on Build GRCh38
rs750680771 Chr.11: 533944-533944 on Build GRCh38
rs730880461 Chr.11: 533853-533853 on Build GRCh38
rs753653328 Chr.11: 533722-533722 on Build GRCh38
rs1049316573 Chr.11: 534086-534086 on Build GRCh38
rs547844807 Chr.11: 534031-534031 on Build GRCh38
rs888029805 Chr.11: 534100-534100 on Build GRCh38
rs587777239 Chr.11: 533869-533870 on Build GRCh38
rs377442252 Chr.11: 533727-533727 on Build GRCh38
rs374666128 Chr.11: 533718-533718 on Build GRCh38
rs747854581 Chr.11: 533757-533757 on Build GRCh38
rs781346394 Chr.11: 533736-533736 on Build GRCh38
rs546681610 Chr.11: 533711-533711 on Build GRCh38
rs375893752 Chr.11: 533774-533774 on Build GRCh38
rs749784082 Chr.11: 533754-533754 on Build GRCh38
rs1013660343 Chr.11: 534116-534116 on Build GRCh38
rs879038333 Chr.11: 534066-534066 on Build GRCh38
rs372223975 Chr.11: 533834-533834 on Build GRCh38
rs749804952 Chr.11: 534071-534071 on Build GRCh38
rs749870259 Chr.11: 533837-533837 on Build GRCh38
rs144718261 Chr.11: 533776-533776 on Build GRCh38
rs45438795 Chr.11: 534044-534045 on Build GRCh38
rs367794954 Chr.11: 533768-533768 on Build GRCh38
rs917139551 Chr.11: 533744-533744 on Build GRCh38
rs778646483 Chr.11: 533731-533731 on Build GRCh38
rs748029060 Chr.11: 533962-533963 on Build GRCh38
rs769056797 Chr.11: 534039-534040 on Build GRCh38
rs898057728 Chr.11: 533861-533861 on Build GRCh38
rs779428004 Chr.11: 533843-533843 on Build GRCh38
rs748524809 Chr.11: 534025-534033 on Build GRCh38
rs771433892 Chr.11: 534076-534076 on Build GRCh38
rs758727504 Chr.11: 533967-533967 on Build GRCh38
rs762706292 Chr.11: 533933-533933 on Build GRCh38
rs1021082161 Chr.11: 534126-534126 on Build GRCh38
rs878854757 Chr.11: 533864-533882 on Build GRCh38
rs1045090499 Chr.11: 534091-534091 on Build GRCh38
rs765074166 Chr.11: 533803-533803 on Build GRCh38
rs775378080 Chr.11: 533930-533930 on Build GRCh38
rs771756228 Chr.11: 533977-533989 on Build GRCh38
rs28933406 Chr.11: 533875-533875 on Build GRCh38
rs954778242 Chr.11: 534040-534041 on Build GRCh38
rs727503092 Chr.11: 533954-533954 on Build GRCh38
rs121913233 Chr.11: 533874-533874 on Build GRCh38
rs756348934 Chr.11: 533951-533951 on Build GRCh38
rs370694089 Chr.11: 534016-534016 on Build GRCh38
rs755286178 Chr.11: 533716-533717 on Build GRCh38
rs1028352031 Chr.11: 533922-533922 on Build GRCh38
rs944477688 Chr.11: 534040-534040 on Build GRCh38
rs977356522 Chr.11: 534022-534022 on Build GRCh38
rs759482455 Chr.11: 533903-533903 on Build GRCh38
rs548634688 Chr.11: 533988-533988 on Build GRCh38
rs769271828 Chr.11: 533993-533993 on Build GRCh38
rs568739285 Chr.11: 533972-533972 on Build GRCh38
rs910272864 Chr.11: 534038-534038 on Build GRCh38
rs770492627 Chr.11: 533884-533884 on Build GRCh38
rs779509554 Chr.11: 533964-533964 on Build GRCh38
rs925794740 Chr.11: 533771-533771 on Build GRCh38
rs767586105 Chr.11: 533819-533819 on Build GRCh38
rs770511125 Chr.11: 533986-533986 on Build GRCh38
rs121913496 Chr.11: 533873-533873 on Build GRCh38
rs776105083 Chr.11: 533885-533885 on Build GRCh38
rs989286439 Chr.11: 534067-534067 on Build GRCh38
rs755488418 Chr.11: 533841-533841 on Build GRCh38
rs763920334 Chr.11: 533936-533936 on Build GRCh38
rs778976132 Chr.11: 533812-533813 on Build GRCh38
rs730880456 Chr.11: 533879-533879 on Build GRCh38
rs587782949 Chr.11: 533779-533779 on Build GRCh38
rs121917758 Chr.11: 533883-533883 on Build GRCh38
rs878854758 Chr.11: 533760-533760 on Build GRCh38
rs753906417 Chr.11: 533958-533958 on Build GRCh38
rs926729312 Chr.11: 533713-533713 on Build GRCh38
rs992164926 Chr.11: 533739-533739 on Build GRCh38
rs765849763 Chr.11: 533798-533798 on Build GRCh38
rs947831654 Chr.11: 534080-534080 on Build GRCh38
rs730880460 Chr.11: 533877-533877 on Build GRCh38
rs945960704 Chr.11: 533746-533746 on Build GRCh38
rs121917756 Chr.11: 533869-533869 on Build GRCh38
rs757406884 Chr.11: 533734-533734 on Build GRCh38
rs1057410548 Chr.11: 534123-534123 on Build GRCh38
rs1002344219 Chr.11: 534102-534102 on Build GRCh38
rs868439179 Chr.11: 533934-533934 on Build GRCh38
rs727504747 Chr.11: 533880-533881 on Build GRCh38
rs749253438 Chr.11: 534084-534085 on Build GRCh38
rs111372582 Chr.11: 533807-533807 on Build GRCh38
rs766582484 Chr.11: 533957-533957 on Build GRCh38
rs727503093 Chr.11: 533881-533881 on Build GRCh38
rs966732125 Chr.11: 534128-534128 on Build GRCh38
rs756190012 Chr.11: 533833-533833 on Build GRCh38
rs745342495 Chr.11: 533989-533989 on Build GRCh38
rs193163027 Chr.11: 533821-533821 on Build GRCh38
rs769212136 Chr.11: 533756-533756 on Build GRCh38
rs935807529 Chr.11: 534032-534032 on Build GRCh38
rs771562113 Chr.11: 533763-533763 on Build GRCh38
rs138272051 Chr.11: 533799-533799 on Build GRCh38
rs927686575 Chr.11: 534092-534092 on Build GRCh38
rs1033725685 Chr.11: 533985-533985 on Build GRCh38
rs1033863305 Chr.11: 534104-534104 on Build GRCh38
rs780757384 Chr.11: 533752-533752 on Build GRCh38
rs533733694 Chr.11: 534130-534130 on Build GRCh38
rs778363504 Chr.11: 534052-534052 on Build GRCh38
rs759463996 Chr.11: 533801-533801 on Build GRCh38
rs1020987166 Chr.11: 534012-534012 on Build GRCh38
rs757505540 Chr.11: 533812-533812 on Build GRCh38
rs557379237 Chr.11: 534037-534037 on Build GRCh38
rs756668252 Chr.11: 533748-533748 on Build GRCh38
rs375166344 Chr.11: 533743-533743 on Build GRCh38
rs746639086 Chr.11: 533882-533882 on Build GRCh38
rs753221782 Chr.11: 533966-533966 on Build GRCh38
rs987229977 Chr.11: 533726-533726 on Build GRCh38
rs1028999343 Chr.11: 534039-534039 on Build GRCh38
rs749674880 Chr.11: 533839-533839 on Build GRCh38

View Full Product Details