Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34357943-34358450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL13
Entrez Gene ID
6357
Gene Name
C-C motif chemokine ligand 13
Gene Aliases
CKb10, MCP-4, NCC-1, NCC1, SCYA13, SCYL1
UniGene
Hs.414629
Gene Chromosome Location
Chr.17: 34356452-34358610 on Build GRCh38
Transcripts
NM_005408.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6032965 c.271C>T p.R91W Chr.17: 34358105-34358105 on Build GRCh38
6146186 c.270C>A p.G90G Chr.17: 34358104-34358104 on Build GRCh38
368646 c.289A>T p.K97* Chr.17: 34358123-34358123 on Build GRCh38
6146187 c.241T>A p.W81R Chr.17: 34358075-34358075 on Build GRCh38
6311308 c.221G>T p.C74F Chr.17: 34358055-34358055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374420615 Chr.17: 34357997-34357997 on Build GRCh38
rs750754428 Chr.17: 34358121-34358128 on Build GRCh38
rs753641013 Chr.17: 34358162-34358162 on Build GRCh38
rs774831219 Chr.17: 34358288-34358289 on Build GRCh38
rs555470907 Chr.17: 34358006-34358006 on Build GRCh38
rs754953835 Chr.17: 34358275-34358275 on Build GRCh38
rs1016404169 Chr.17: 34358102-34358102 on Build GRCh38
rs1022291677 Chr.17: 34358375-34358375 on Build GRCh38
rs137918947 Chr.17: 34358073-34358073 on Build GRCh38
rs2072069 Chr.17: 34357972-34357972 on Build GRCh38
rs777206868 Chr.17: 34358183-34358183 on Build GRCh38
rs757885510 Chr.17: 34358110-34358110 on Build GRCh38
rs533284969 Chr.17: 34358424-34358424 on Build GRCh38
rs896894814 Chr.17: 34358306-34358306 on Build GRCh38
rs926180063 Chr.17: 34358241-34358241 on Build GRCh38
rs771895967 Chr.17: 34358217-34358217 on Build GRCh38
rs763086648 Chr.17: 34358139-34358139 on Build GRCh38
rs183253878 Chr.17: 34358105-34358105 on Build GRCh38
rs766223344 Chr.17: 34358079-34358079 on Build GRCh38
rs776910644 Chr.17: 34358112-34358112 on Build GRCh38
rs867324342 Chr.17: 34358226-34358226 on Build GRCh38
rs143510106 Chr.17: 34358041-34358041 on Build GRCh38
rs1043601675 Chr.17: 34358237-34358237 on Build GRCh38
rs778740838 Chr.17: 34358104-34358104 on Build GRCh38
rs1026848084 Chr.17: 34358048-34358048 on Build GRCh38
rs989835234 Chr.17: 34358369-34358369 on Build GRCh38
rs752642615 Chr.17: 34358174-34358174 on Build GRCh38
rs1028776595 Chr.17: 34358143-34358143 on Build GRCh38
rs762063628 Chr.17: 34358142-34358142 on Build GRCh38
rs187454526 Chr.17: 34358106-34358106 on Build GRCh38
rs993972988 Chr.17: 34358011-34358011 on Build GRCh38
rs372653174 Chr.17: 34358111-34358111 on Build GRCh38
rs528475517 Chr.17: 34358250-34358250 on Build GRCh38
rs993955380 Chr.17: 34358335-34358335 on Build GRCh38
rs149499630 Chr.17: 34358357-34358357 on Build GRCh38
rs373186520 Chr.17: 34358377-34358377 on Build GRCh38
rs755965057 Chr.17: 34358077-34358077 on Build GRCh38
rs777108695 Chr.17: 34358020-34358020 on Build GRCh38
rs75348820 Chr.17: 34358428-34358428 on Build GRCh38
rs749455119 Chr.17: 34358278-34358278 on Build GRCh38
rs191543093 Chr.17: 34358219-34358219 on Build GRCh38
rs774589882 Chr.17: 34358141-34358141 on Build GRCh38
rs542740740 Chr.17: 34358201-34358201 on Build GRCh38
rs898861282 Chr.17: 34358307-34358307 on Build GRCh38
rs201259871 Chr.17: 34357988-34357988 on Build GRCh38
rs757912599 Chr.17: 34358181-34358181 on Build GRCh38
rs1056015838 Chr.17: 34358273-34358273 on Build GRCh38
rs146257516 Chr.17: 34357980-34357980 on Build GRCh38
rs940311334 Chr.17: 34358275-34358276 on Build GRCh38
rs759975138 Chr.17: 34358037-34358037 on Build GRCh38
rs764140711 Chr.17: 34358140-34358140 on Build GRCh38
rs771707528 Chr.17: 34358004-34358004 on Build GRCh38
rs980838816 Chr.17: 34358382-34358382 on Build GRCh38
rs976742176 Chr.17: 34358114-34358114 on Build GRCh38
rs762960516 Chr.17: 34358050-34358050 on Build GRCh38
rs1018125310 Chr.17: 34358372-34358372 on Build GRCh38
rs377174787 Chr.17: 34358159-34358159 on Build GRCh38
rs534660128 Chr.17: 34358055-34358055 on Build GRCh38
rs969815975 Chr.17: 34358380-34358380 on Build GRCh38
rs965310874 Chr.17: 34358097-34358097 on Build GRCh38
rs750055494 Chr.17: 34358015-34358015 on Build GRCh38
rs868196051 Chr.17: 34358360-34358360 on Build GRCh38
rs557754223 Chr.17: 34358274-34358274 on Build GRCh38
rs573925061 Chr.17: 34358129-34358129 on Build GRCh38
rs751463485 Chr.17: 34358166-34358166 on Build GRCh38
rs915275556 Chr.17: 34358189-34358189 on Build GRCh38
rs749298428 Chr.17: 34357995-34357995 on Build GRCh38
rs777705769 Chr.17: 34358277-34358277 on Build GRCh38
rs577924843 Chr.17: 34358085-34358085 on Build GRCh38
rs371169143 Chr.17: 34357996-34357996 on Build GRCh38
rs370593553 Chr.17: 34358167-34358167 on Build GRCh38
rs771349142 Chr.17: 34358013-34358013 on Build GRCh38
rs758137997 Chr.17: 34358051-34358051 on Build GRCh38
rs1030854868 Chr.17: 34358359-34358359 on Build GRCh38
rs749651505 Chr.17: 34358089-34358089 on Build GRCh38
rs890276031 Chr.17: 34358343-34358343 on Build GRCh38
rs534656266 Chr.17: 34358000-34358000 on Build GRCh38
rs754905287 Chr.17: 34358098-34358098 on Build GRCh38
rs1028724162 Chr.17: 34358339-34358339 on Build GRCh38
rs377265823 Chr.17: 34358001-34358001 on Build GRCh38
rs775907825 Chr.17: 34358137-34358137 on Build GRCh38
rs139142355 Chr.17: 34358336-34358336 on Build GRCh38
rs367613739 Chr.17: 34358043-34358043 on Build GRCh38
rs779852870 Chr.17: 34358133-34358133 on Build GRCh38
rs948152399 Chr.17: 34358205-34358205 on Build GRCh38
rs1046108785 Chr.17: 34358295-34358295 on Build GRCh38

View Full Product Details