Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34358328-34358841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL13
Entrez Gene ID
6357
Gene Name
C-C motif chemokine ligand 13
Gene Aliases
CKb10, MCP-4, NCC-1, NCC1, SCYA13, SCYL1
UniGene
Hs.414629
Gene Chromosome Location
Chr.17: 34356452-34358610 on Build GRCh38
Transcripts
NM_005408.2

SNP Detail

SNP ID Chromosome Location
rs534280819 Chr.17: 34358559-34358559 on Build GRCh38
rs917476487 Chr.17: 34358499-34358499 on Build GRCh38
rs1010804243 Chr.17: 34358595-34358595 on Build GRCh38
rs929639588 Chr.17: 34358541-34358541 on Build GRCh38
rs565640851 Chr.17: 34358780-34358780 on Build GRCh38
rs941614103 Chr.17: 34358467-34358467 on Build GRCh38
rs901081101 Chr.17: 34358609-34358609 on Build GRCh38
rs1022291677 Chr.17: 34358375-34358375 on Build GRCh38
rs1006698136 Chr.17: 34358580-34358580 on Build GRCh38
rs148683606 Chr.17: 34358684-34358684 on Build GRCh38
rs533284969 Chr.17: 34358424-34358424 on Build GRCh38
rs929635681 Chr.17: 34358777-34358777 on Build GRCh38
rs537319630 Chr.17: 34358701-34358701 on Build GRCh38
rs896859550 Chr.17: 34358524-34358524 on Build GRCh38
rs1035063946 Chr.17: 34358667-34358667 on Build GRCh38
rs186581102 Chr.17: 34358504-34358504 on Build GRCh38
rs543374329 Chr.17: 34358630-34358630 on Build GRCh38
rs776037462 Chr.17: 34358510-34358510 on Build GRCh38
rs978901702 Chr.17: 34358446-34358446 on Build GRCh38
rs989835234 Chr.17: 34358369-34358369 on Build GRCh38
rs892285032 Chr.17: 34358800-34358800 on Build GRCh38
rs1018094335 Chr.17: 34358592-34358592 on Build GRCh38
rs982898798 Chr.17: 34358677-34358677 on Build GRCh38
rs149499630 Chr.17: 34358357-34358357 on Build GRCh38
rs373186520 Chr.17: 34358377-34358377 on Build GRCh38
rs75348820 Chr.17: 34358428-34358428 on Build GRCh38
rs1050107902 Chr.17: 34358521-34358521 on Build GRCh38
rs772526231 Chr.17: 34358489-34358489 on Build GRCh38
rs867452490 Chr.17: 34358512-34358512 on Build GRCh38
rs1046078642 Chr.17: 34358515-34358515 on Build GRCh38
rs942470183 Chr.17: 34358808-34358808 on Build GRCh38
rs185659231 Chr.17: 34358565-34358565 on Build GRCh38
rs908707040 Chr.17: 34358678-34358678 on Build GRCh38
rs931627352 Chr.17: 34358726-34358726 on Build GRCh38
rs937484917 Chr.17: 34358458-34358458 on Build GRCh38
rs11575023 Chr.17: 34358724-34358724 on Build GRCh38
rs980838816 Chr.17: 34358382-34358382 on Build GRCh38
rs543207368 Chr.17: 34358748-34358748 on Build GRCh38
rs1018125310 Chr.17: 34358372-34358372 on Build GRCh38
rs982950949 Chr.17: 34358434-34358434 on Build GRCh38
rs969815975 Chr.17: 34358380-34358380 on Build GRCh38
rs192522491 Chr.17: 34358556-34358556 on Build GRCh38
rs777007300 Chr.17: 34358807-34358807 on Build GRCh38
rs868196051 Chr.17: 34358360-34358360 on Build GRCh38
rs892330250 Chr.17: 34358589-34358589 on Build GRCh38
rs374778955 Chr.17: 34358654-34358654 on Build GRCh38
rs960788282 Chr.17: 34358676-34358676 on Build GRCh38
rs1033072819 Chr.17: 34358699-34358699 on Build GRCh38
rs144148427 Chr.17: 34358431-34358431 on Build GRCh38
rs926036948 Chr.17: 34358456-34358456 on Build GRCh38
rs1048549686 Chr.17: 34358806-34358806 on Build GRCh38
rs1052635204 Chr.17: 34358799-34358799 on Build GRCh38
rs998099977 Chr.17: 34358610-34358610 on Build GRCh38
rs145360158 Chr.17: 34358738-34358738 on Build GRCh38
rs573890586 Chr.17: 34358750-34358758 on Build GRCh38
rs950277268 Chr.17: 34358732-34358732 on Build GRCh38
rs746116756 Chr.17: 34358444-34358444 on Build GRCh38
rs1030854868 Chr.17: 34358359-34358359 on Build GRCh38
rs78322034 Chr.17: 34358731-34358731 on Build GRCh38
rs890223863 Chr.17: 34358534-34358534 on Build GRCh38
rs1020232351 Chr.17: 34358625-34358625 on Build GRCh38
rs920256881 Chr.17: 34358497-34358497 on Build GRCh38
rs950307999 Chr.17: 34358500-34358500 on Build GRCh38
rs535679541 Chr.17: 34358432-34358432 on Build GRCh38
rs919512952 Chr.17: 34358754-34358754 on Build GRCh38
rs182653739 Chr.17: 34358452-34358452 on Build GRCh38

View Full Product Details