Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35976876-35977408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL16
Entrez Gene ID
6360
Gene Name
C-C motif chemokine ligand 16
Gene Aliases
CKb12, HCC-4, ILINCK, LCC-1, LEC, LMC, Mtn-1, NCC-4, NCC4, SCYA16, SCYL4
UniGene
Hs.10458
Gene Chromosome Location
Chr.17: 35976493-35983620 on Build GRCh38
Transcripts
NM_004590.3 XM_005258020.4

SNP Detail

SNP ID Chromosome Location
rs1023445602 Chr.17: 35977187-35977187 on Build GRCh38
rs1052467158 Chr.17: 35977094-35977094 on Build GRCh38
rs1049467200 Chr.17: 35977378-35977378 on Build GRCh38
rs555046042 Chr.17: 35977034-35977034 on Build GRCh38
rs974825986 Chr.17: 35977342-35977342 on Build GRCh38
rs956117092 Chr.17: 35977177-35977177 on Build GRCh38
rs184264470 Chr.17: 35977316-35977316 on Build GRCh38
rs1037069776 Chr.17: 35977042-35977042 on Build GRCh38
rs865815886 Chr.17: 35977314-35977314 on Build GRCh38
rs979530991 Chr.17: 35976921-35976921 on Build GRCh38
rs192123970 Chr.17: 35977103-35977103 on Build GRCh38
rs983825822 Chr.17: 35977284-35977284 on Build GRCh38
rs1734967 Chr.17: 35976945-35976945 on Build GRCh38
rs950801096 Chr.17: 35977007-35977007 on Build GRCh38
rs926749097 Chr.17: 35976931-35976931 on Build GRCh38
rs963407925 Chr.17: 35977340-35977340 on Build GRCh38
rs1026021927 Chr.17: 35977214-35977214 on Build GRCh38
rs868434826 Chr.17: 35977002-35977002 on Build GRCh38
rs1012019998 Chr.17: 35977186-35977186 on Build GRCh38
rs191970638 Chr.17: 35977247-35977247 on Build GRCh38
rs183338583 Chr.17: 35977121-35977121 on Build GRCh38
rs551618958 Chr.17: 35977000-35977000 on Build GRCh38
rs922083234 Chr.17: 35977358-35977358 on Build GRCh38
rs940226359 Chr.17: 35977318-35977318 on Build GRCh38
rs557313309 Chr.17: 35977038-35977038 on Build GRCh38
rs938156932 Chr.17: 35977291-35977291 on Build GRCh38
rs988748272 Chr.17: 35977210-35977210 on Build GRCh38
rs980966622 Chr.17: 35976954-35976954 on Build GRCh38
rs187547039 Chr.17: 35977146-35977146 on Build GRCh38
rs529366348 Chr.17: 35977357-35977357 on Build GRCh38
rs933559408 Chr.17: 35977072-35977072 on Build GRCh38
rs987730569 Chr.17: 35977367-35977367 on Build GRCh38
rs935599098 Chr.17: 35977383-35977383 on Build GRCh38
rs928640914 Chr.17: 35976959-35976959 on Build GRCh38
rs187777672 Chr.17: 35977031-35977031 on Build GRCh38
rs1039140225 Chr.17: 35977045-35977045 on Build GRCh38
rs571551460 Chr.17: 35977382-35977382 on Build GRCh38
rs549700470 Chr.17: 35977369-35977369 on Build GRCh38
rs759426136 Chr.17: 35976961-35976961 on Build GRCh38
rs906394050 Chr.17: 35977197-35977197 on Build GRCh38
rs544382679 Chr.17: 35977306-35977306 on Build GRCh38
rs1127790 Chr.17: 35977211-35977211 on Build GRCh38
rs992769442 Chr.17: 35977309-35977309 on Build GRCh38
rs922114848 Chr.17: 35977054-35977054 on Build GRCh38
rs898160492 Chr.17: 35977052-35977052 on Build GRCh38
rs150951362 Chr.17: 35977228-35977228 on Build GRCh38
rs60941529 Chr.17: 35977159-35977159 on Build GRCh38
rs933527131 Chr.17: 35977363-35977363 on Build GRCh38
rs982038982 Chr.17: 35976898-35976898 on Build GRCh38
rs144545525 Chr.17: 35977259-35977273 on Build GRCh38

View Full Product Details