Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35978057-35978560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL16
Entrez Gene ID
6360
Gene Name
C-C motif chemokine ligand 16
Gene Aliases
CKb12, HCC-4, ILINCK, LCC-1, LEC, LMC, Mtn-1, NCC-4, NCC4, SCYA16, SCYL4
UniGene
Hs.10458
Gene Chromosome Location
Chr.17: 35976493-35983620 on Build GRCh38
Transcripts
NM_004590.3 XM_005258020.4

SNP Detail

SNP ID Chromosome Location
rs545074841 Chr.17: 35978359-35978359 on Build GRCh38
rs1031125572 Chr.17: 35978361-35978361 on Build GRCh38
rs917015 Chr.17: 35978489-35978489 on Build GRCh38
rs143888448 Chr.17: 35978223-35978223 on Build GRCh38
rs765375095 Chr.17: 35978190-35978190 on Build GRCh38
rs773867999 Chr.17: 35978157-35978157 on Build GRCh38
rs899720236 Chr.17: 35978257-35978257 on Build GRCh38
rs761608144 Chr.17: 35978208-35978208 on Build GRCh38
rs375803541 Chr.17: 35978249-35978249 on Build GRCh38
rs895768902 Chr.17: 35978526-35978526 on Build GRCh38
rs201371224 Chr.17: 35978278-35978278 on Build GRCh38
rs111316280 Chr.17: 35978236-35978236 on Build GRCh38
rs373312061 Chr.17: 35978097-35978097 on Build GRCh38
rs908249318 Chr.17: 35978513-35978513 on Build GRCh38
rs182157112 Chr.17: 35978164-35978164 on Build GRCh38
rs748995993 Chr.17: 35978135-35978135 on Build GRCh38
rs747780883 Chr.17: 35978311-35978311 on Build GRCh38
rs755992491 Chr.17: 35978240-35978240 on Build GRCh38
rs773058070 Chr.17: 35978514-35978514 on Build GRCh38
rs985664596 Chr.17: 35978323-35978323 on Build GRCh38
rs370055535 Chr.17: 35978124-35978124 on Build GRCh38
rs11080369 Chr.17: 35978128-35978128 on Build GRCh38
rs757286891 Chr.17: 35978100-35978100 on Build GRCh38
rs114853983 Chr.17: 35978326-35978326 on Build GRCh38
rs754729739 Chr.17: 35978293-35978293 on Build GRCh38
rs772761377 Chr.17: 35978153-35978153 on Build GRCh38
rs982929421 Chr.17: 35978511-35978511 on Build GRCh38
rs981035205 Chr.17: 35978481-35978481 on Build GRCh38
rs574724630 Chr.17: 35978187-35978187 on Build GRCh38
rs375916625 Chr.17: 35978134-35978134 on Build GRCh38
rs150085057 Chr.17: 35978536-35978536 on Build GRCh38
rs530456077 Chr.17: 35978515-35978515 on Build GRCh38
rs778419997 Chr.17: 35978309-35978309 on Build GRCh38
rs765967537 Chr.17: 35978248-35978248 on Build GRCh38
rs890489589 Chr.17: 35978336-35978336 on Build GRCh38
rs952896410 Chr.17: 35978297-35978297 on Build GRCh38
rs565397401 Chr.17: 35978369-35978369 on Build GRCh38
rs780155359 Chr.17: 35978166-35978166 on Build GRCh38
rs965414174 Chr.17: 35978327-35978327 on Build GRCh38
rs776524378 Chr.17: 35978184-35978184 on Build GRCh38
rs1046832750 Chr.17: 35978151-35978151 on Build GRCh38
rs544599289 Chr.17: 35978473-35978473 on Build GRCh38
rs771166279 Chr.17: 35978169-35978169 on Build GRCh38
rs1011045364 Chr.17: 35978434-35978434 on Build GRCh38
rs537862229 Chr.17: 35978149-35978149 on Build GRCh38
rs945776421 Chr.17: 35978394-35978394 on Build GRCh38
rs915531019 Chr.17: 35978113-35978113 on Build GRCh38
rs937184947 Chr.17: 35978494-35978494 on Build GRCh38
rs969607390 Chr.17: 35978451-35978451 on Build GRCh38
rs941235368 Chr.17: 35978183-35978183 on Build GRCh38
rs368258545 Chr.17: 35978207-35978207 on Build GRCh38
rs755628216 Chr.17: 35978133-35978133 on Build GRCh38
rs767230651 Chr.17: 35978219-35978219 on Build GRCh38
rs754560966 Chr.17: 35978142-35978142 on Build GRCh38
rs770190565 Chr.17: 35978131-35978136 on Build GRCh38
rs960441577 Chr.17: 35978490-35978490 on Build GRCh38
rs552180846 Chr.17: 35978099-35978099 on Build GRCh38
rs140793906 Chr.17: 35978188-35978188 on Build GRCh38
rs575510758 Chr.17: 35978450-35978450 on Build GRCh38
rs748454851 Chr.17: 35978150-35978150 on Build GRCh38

View Full Product Details