Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17825069-17825596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
UniGene
Hs.515247
Gene Chromosome Location
Chr.19: 17824782-17848071 on Build GRCh38
Transcripts
NM_000215.3 XM_005259896.3

SNP Detail

SNP ID Chromosome Location
rs886054268 Chr.19: 17825212-17825212 on Build GRCh38
rs935679583 Chr.19: 17825504-17825504 on Build GRCh38
rs771793558 Chr.19: 17825456-17825456 on Build GRCh38
rs73014967 Chr.19: 17825450-17825450 on Build GRCh38
rs919233223 Chr.19: 17825129-17825129 on Build GRCh38
rs891046335 Chr.19: 17825400-17825400 on Build GRCh38
rs561294941 Chr.19: 17825380-17825380 on Build GRCh38
rs752515408 Chr.19: 17825363-17825363 on Build GRCh38
rs564743916 Chr.19: 17825308-17825308 on Build GRCh38
rs749028771 Chr.19: 17825333-17825333 on Build GRCh38
rs886054269 Chr.19: 17825389-17825390 on Build GRCh38
rs909732304 Chr.19: 17825153-17825153 on Build GRCh38
rs950010878 Chr.19: 17825477-17825477 on Build GRCh38
rs145985086 Chr.19: 17825220-17825220 on Build GRCh38
rs1006802984 Chr.19: 17825414-17825414 on Build GRCh38
rs1055535802 Chr.19: 17825163-17825163 on Build GRCh38
rs966705186 Chr.19: 17825255-17825255 on Build GRCh38
rs548595316 Chr.19: 17825567-17825567 on Build GRCh38
rs533856149 Chr.19: 17825137-17825137 on Build GRCh38
rs1011325577 Chr.19: 17825174-17825174 on Build GRCh38
rs549996792 Chr.19: 17825431-17825431 on Build GRCh38
rs545062692 Chr.19: 17825286-17825286 on Build GRCh38
rs534968482 Chr.19: 17825551-17825551 on Build GRCh38
rs542886257 Chr.19: 17825183-17825183 on Build GRCh38
rs3213415 Chr.19: 17825546-17825546 on Build GRCh38
rs560941492 Chr.19: 17825134-17825134 on Build GRCh38
rs553990627 Chr.19: 17825143-17825143 on Build GRCh38
rs938095902 Chr.19: 17825447-17825447 on Build GRCh38
rs191522931 Chr.19: 17825195-17825195 on Build GRCh38
rs183431894 Chr.19: 17825436-17825436 on Build GRCh38
rs1032730913 Chr.19: 17825212-17825212 on Build GRCh38
rs755967027 Chr.19: 17825278-17825278 on Build GRCh38
rs7508719 Chr.19: 17825513-17825513 on Build GRCh38
rs552602675 Chr.19: 17825479-17825479 on Build GRCh38
rs118101939 Chr.19: 17825424-17825424 on Build GRCh38
rs984743375 Chr.19: 17825330-17825330 on Build GRCh38
rs937204492 Chr.19: 17825155-17825155 on Build GRCh38
rs994168731 Chr.19: 17825572-17825572 on Build GRCh38
rs992101375 Chr.19: 17825239-17825239 on Build GRCh38
rs1019117117 Chr.19: 17825246-17825246 on Build GRCh38
rs902858566 Chr.19: 17825502-17825502 on Build GRCh38
rs1018074991 Chr.19: 17825151-17825151 on Build GRCh38
rs79972191 Chr.19: 17825321-17825321 on Build GRCh38
rs1007140432 Chr.19: 17825113-17825115 on Build GRCh38
rs528896845 Chr.19: 17825387-17825387 on Build GRCh38
rs541364775 Chr.19: 17825329-17825329 on Build GRCh38
rs1019128591 Chr.19: 17825541-17825541 on Build GRCh38
rs958950984 Chr.19: 17825236-17825236 on Build GRCh38
rs928891616 Chr.19: 17825232-17825232 on Build GRCh38
rs1035345922 Chr.19: 17825522-17825522 on Build GRCh38
rs754759025 Chr.19: 17825382-17825382 on Build GRCh38
rs895775234 Chr.19: 17825169-17825169 on Build GRCh38
rs538564066 Chr.19: 17825505-17825505 on Build GRCh38
rs750558618 Chr.19: 17825234-17825234 on Build GRCh38
rs772556293 Chr.19: 17825521-17825521 on Build GRCh38
rs886054270 Chr.19: 17825525-17825525 on Build GRCh38
rs949988587 Chr.19: 17825193-17825193 on Build GRCh38
rs573881908 Chr.19: 17825161-17825161 on Build GRCh38
rs888906584 Chr.19: 17825514-17825514 on Build GRCh38
rs942131845 Chr.19: 17825256-17825256 on Build GRCh38
rs902808842 Chr.19: 17825208-17825208 on Build GRCh38
rs757613147 Chr.19: 17825111-17825111 on Build GRCh38
rs966300855 Chr.19: 17825104-17825104 on Build GRCh38
rs972509380 Chr.19: 17825106-17825106 on Build GRCh38
rs972304989 Chr.19: 17825291-17825291 on Build GRCh38
rs1000368007 Chr.19: 17825210-17825210 on Build GRCh38

View Full Product Details