Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28015570-28015727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT3
Entrez Gene ID
2322
Gene Name
fms related tyrosine kinase 3
Gene Aliases
CD135, FLK-2, FLK2, STK1
UniGene
Hs.507590
Gene Chromosome Location
Chr.13: 28003274-28100592 on Build GRCh38
Transcripts
NM_004119.2 NR_130706.1 XM_011535015.2 XM_011535017.2 XM_011535018.2 XM_017020486.1 XM_017020487.1 XM_017020488.1 XM_017020489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5786093 c.2578C>A p.L860M Chr.13: 28015665-28015665 on Build GRCh38
6138847 c.2648C>A p.S883* Chr.13: 28015595-28015595 on Build GRCh38
4538943 c.2623G>A p.G875R Chr.13: 28015620-28015620 on Build GRCh38
416399 c.2634G>A p.L878L Chr.13: 28015609-28015609 on Build GRCh38
6947067 c.2651T>C p.L884P Chr.13: 28015592-28015592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959896930 Chr.13: 28015698-28015698 on Build GRCh38
rs754418964 Chr.13: 28015693-28015693 on Build GRCh38
rs772553557 Chr.13: 28015630-28015630 on Build GRCh38
rs771026522 Chr.13: 28015645-28015645 on Build GRCh38
rs868802040 Chr.13: 28015622-28015622 on Build GRCh38
rs201398123 Chr.13: 28015647-28015647 on Build GRCh38
rs750047857 Chr.13: 28015651-28015651 on Build GRCh38
rs762000524 Chr.13: 28015621-28015621 on Build GRCh38
rs903856095 Chr.13: 28015667-28015667 on Build GRCh38
rs201923726 Chr.13: 28015697-28015697 on Build GRCh38
rs774360726 Chr.13: 28015664-28015664 on Build GRCh38
rs773272572 Chr.13: 28015676-28015676 on Build GRCh38
rs768723625 Chr.13: 28015660-28015660 on Build GRCh38
rs753361791 Chr.13: 28015689-28015689 on Build GRCh38
rs968671409 Chr.13: 28015642-28015642 on Build GRCh38
rs769918229 Chr.13: 28015666-28015666 on Build GRCh38
rs753753892 Chr.13: 28015653-28015653 on Build GRCh38
rs927209673 Chr.13: 28015696-28015696 on Build GRCh38
rs368432815 Chr.13: 28015633-28015633 on Build GRCh38
rs573768667 Chr.13: 28015675-28015675 on Build GRCh38
rs370459694 Chr.13: 28015601-28015601 on Build GRCh38
rs367899009 Chr.13: 28015600-28015600 on Build GRCh38
rs568745490 Chr.13: 28015671-28015671 on Build GRCh38
rs1050822599 Chr.13: 28015665-28015665 on Build GRCh38
rs772854825 Chr.13: 28015672-28015672 on Build GRCh38
rs776952835 Chr.13: 28015604-28015604 on Build GRCh38
rs147075807 Chr.13: 28015687-28015687 on Build GRCh38
rs200175703 Chr.13: 28015588-28015588 on Build GRCh38
rs779336061 Chr.13: 28015609-28015609 on Build GRCh38
rs759638195 Chr.13: 28015649-28015649 on Build GRCh38

View Full Product Details