Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 164792401-164792574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PBX1
Entrez Gene ID
5087
Gene Name
PBX homeobox 1
Gene Aliases
-
UniGene
Hs.557097
Gene Chromosome Location
Chr.1: 164559360-164886047 on Build GRCh38
Transcripts
NM_001204961.1 NM_001204963.1 NM_002585.3 XM_005245228.3 XM_005245229.3 XM_011509590.1 XM_011509591.2 XM_011509592.1 XM_017001395.1 XM_017001396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1689064 c.295G>A p.E99K Chr.1: 164792523-164792523 on Build GRCh38
3863385 c.286C>T p.Q96* Chr.1: 164792514-164792514 on Build GRCh38
6005821 c.340G>T p.A114S Chr.1: 164792568-164792568 on Build GRCh38
4224711 c.278G>A p.R93Q Chr.1: 164792506-164792506 on Build GRCh38
6738438 c.5G>A p.R2Q Chr.1: 164792548-164792548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749609233 Chr.1: 164792480-164792480 on Build GRCh38
rs1002668836 Chr.1: 164792471-164792471 on Build GRCh38
rs377380016 Chr.1: 164792455-164792455 on Build GRCh38
rs757647151 Chr.1: 164792553-164792553 on Build GRCh38
rs750091180 Chr.1: 164792447-164792447 on Build GRCh38
rs757803669 Chr.1: 164792454-164792454 on Build GRCh38
rs35543782 Chr.1: 164792509-164792509 on Build GRCh38
rs778272148 Chr.1: 164792462-164792462 on Build GRCh38
rs756755109 Chr.1: 164792456-164792456 on Build GRCh38
rs139680846 Chr.1: 164792478-164792479 on Build GRCh38
rs867567117 Chr.1: 164792517-164792517 on Build GRCh38

View Full Product Details