Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159993728-159993967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
UniGene
Hs.673
Gene Chromosome Location
Chr.3: 159988836-159996019 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Gene Symbol
IL12A-AS1
Entrez Gene ID
101928376
Gene Name
IL12A antisense RNA 1
Gene Aliases
-
UniGene
Hs.565865
Gene Chromosome Location
Chr.3: 159913400-160207092 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3589426 c.554G>A p.R185K Chr.3: 159993792-159993792 on Build GRCh38
1617136 c.559A>G p.I187V Chr.3: 159993797-159993797 on Build GRCh38
3005966 c.552G>T p.K184N Chr.3: 159993790-159993790 on Build GRCh38
6163673 c.600G>T p.L200L Chr.3: 159993838-159993838 on Build GRCh38
3005965 c.507G>C p.Q169H Chr.3: 159993745-159993745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749886054 Chr.3: 159993882-159993882 on Build GRCh38
rs773136641 Chr.3: 159993806-159993806 on Build GRCh38
rs773148606 Chr.3: 159993823-159993823 on Build GRCh38
rs535488207 Chr.3: 159993905-159993906 on Build GRCh38
rs765267336 Chr.3: 159993875-159993875 on Build GRCh38
rs2230052 Chr.3: 159993805-159993805 on Build GRCh38
rs369508050 Chr.3: 159993870-159993870 on Build GRCh38
rs748559121 Chr.3: 159993791-159993791 on Build GRCh38
rs935618466 Chr.3: 159993880-159993880 on Build GRCh38
rs76693142 Chr.3: 159993921-159993921 on Build GRCh38
rs1024513737 Chr.3: 159993815-159993815 on Build GRCh38
rs988377448 Chr.3: 159993903-159993903 on Build GRCh38
rs538084986 Chr.3: 159993843-159993843 on Build GRCh38
rs781551474 Chr.3: 159993764-159993764 on Build GRCh38
rs545507796 Chr.3: 159993905-159993905 on Build GRCh38
rs763929222 Chr.3: 159993852-159993852 on Build GRCh38
rs556606966 Chr.3: 159993844-159993844 on Build GRCh38
rs574173919 Chr.3: 159993810-159993810 on Build GRCh38
rs964398253 Chr.3: 159993833-159993833 on Build GRCh38
rs1001624071 Chr.3: 159993827-159993827 on Build GRCh38
rs976811495 Chr.3: 159993861-159993861 on Build GRCh38
rs779545994 Chr.3: 159993885-159993885 on Build GRCh38
rs753716045 Chr.3: 159993860-159993860 on Build GRCh38
rs768944534 Chr.3: 159993760-159993760 on Build GRCh38
rs201524139 Chr.3: 159993883-159993883 on Build GRCh38
rs754648444 Chr.3: 159993895-159993895 on Build GRCh38
rs55780930 Chr.3: 159993786-159993786 on Build GRCh38
rs751154856 Chr.3: 159993894-159993894 on Build GRCh38

View Full Product Details