Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35981240-35981513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL16
Entrez Gene ID
6360
Gene Name
C-C motif chemokine ligand 16
Gene Aliases
CKb12, HCC-4, ILINCK, LCC-1, LEC, LMC, Mtn-1, NCC-4, NCC4, SCYA16, SCYL4
UniGene
Hs.10458
Gene Chromosome Location
Chr.17: 35976493-35983620 on Build GRCh38
Transcripts
NM_004590.3 XM_005258020.4

SNP Detail

SNP ID Chromosome Location
rs772965529 Chr.17: 35981321-35981321 on Build GRCh38
rs753448636 Chr.17: 35981335-35981335 on Build GRCh38
rs199602184 Chr.17: 35981365-35981365 on Build GRCh38
rs201562530 Chr.17: 35981434-35981434 on Build GRCh38
rs771195688 Chr.17: 35981303-35981303 on Build GRCh38
rs549950289 Chr.17: 35981487-35981487 on Build GRCh38
rs543053536 Chr.17: 35981356-35981356 on Build GRCh38
rs369188602 Chr.17: 35981440-35981440 on Build GRCh38
rs930541798 Chr.17: 35981473-35981473 on Build GRCh38
rs752429863 Chr.17: 35981357-35981357 on Build GRCh38
rs769792851 Chr.17: 35981375-35981375 on Build GRCh38
rs372500078 Chr.17: 35981313-35981313 on Build GRCh38
rs768840174 Chr.17: 35981387-35981387 on Build GRCh38
rs996850914 Chr.17: 35981474-35981474 on Build GRCh38
rs1032661925 Chr.17: 35981374-35981374 on Build GRCh38
rs757695950 Chr.17: 35981414-35981414 on Build GRCh38
rs758075390 Chr.17: 35981358-35981358 on Build GRCh38
rs370263354 Chr.17: 35981429-35981429 on Build GRCh38
rs987641482 Chr.17: 35981310-35981310 on Build GRCh38
rs919117996 Chr.17: 35981467-35981467 on Build GRCh38
rs141803122 Chr.17: 35981384-35981384 on Build GRCh38
rs759711291 Chr.17: 35981472-35981472 on Build GRCh38
rs760453337 Chr.17: 35981405-35981405 on Build GRCh38
rs1049005670 Chr.17: 35981486-35981486 on Build GRCh38
rs774573988 Chr.17: 35981465-35981467 on Build GRCh38
rs148292019 Chr.17: 35981380-35981380 on Build GRCh38
rs933061994 Chr.17: 35981276-35981276 on Build GRCh38
rs116518274 Chr.17: 35981364-35981364 on Build GRCh38
rs759479400 Chr.17: 35981409-35981409 on Build GRCh38
rs781562264 Chr.17: 35981368-35981368 on Build GRCh38
rs754164622 Chr.17: 35981263-35981263 on Build GRCh38
rs116077102 Chr.17: 35981456-35981456 on Build GRCh38
rs770704126 Chr.17: 35981406-35981406 on Build GRCh38
rs763824864 Chr.17: 35981433-35981433 on Build GRCh38
rs200938478 Chr.17: 35981314-35981314 on Build GRCh38
rs774388713 Chr.17: 35981466-35981467 on Build GRCh38
rs764676027 Chr.17: 35981426-35981426 on Build GRCh38
rs577525020 Chr.17: 35981337-35981337 on Build GRCh38
rs760649743 Chr.17: 35981326-35981326 on Build GRCh38
rs979038372 Chr.17: 35981370-35981370 on Build GRCh38
rs542591738 Chr.17: 35981430-35981430 on Build GRCh38
rs1044362941 Chr.17: 35981412-35981412 on Build GRCh38
rs563594361 Chr.17: 35981362-35981362 on Build GRCh38
rs376685257 Chr.17: 35981437-35981437 on Build GRCh38
rs769851900 Chr.17: 35981306-35981306 on Build GRCh38
rs368916906 Chr.17: 35981392-35981392 on Build GRCh38
rs750913925 Chr.17: 35981442-35981442 on Build GRCh38
rs369776507 Chr.17: 35981262-35981262 on Build GRCh38
rs142115594 Chr.17: 35981408-35981408 on Build GRCh38
rs746318032 Chr.17: 35981369-35981369 on Build GRCh38
rs371857407 Chr.17: 35981455-35981455 on Build GRCh38
rs766225384 Chr.17: 35981332-35981332 on Build GRCh38
rs745921070 Chr.17: 35981300-35981300 on Build GRCh38

View Full Product Details