Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243505166-243505439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT3
Entrez Gene ID
10000
Gene Name
AKT serine/threonine kinase 3
Gene Aliases
MPPH, MPPH2, PKB-GAMMA, PKBG, PRKBG, RAC-PK-gamma, RAC-gamma, STK-2
UniGene
Hs.498292
Gene Chromosome Location
Chr.1: 243488233-243851079 on Build GRCh38
Transcripts
NM_001206729.1 NM_005465.4 NM_181690.2 XM_005272994.4 XM_005272995.2 XM_011544014.2 XM_016999985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6960375 c.1432C>T p.R478* Chr.1: 243505257-243505257 on Build GRCh38
6045404 c.1398G>A p.P466P Chr.1: 243505291-243505291 on Build GRCh38
414540 c.1410A>G p.Q470Q Chr.1: 243505279-243505279 on Build GRCh38
4165115 c.1425A>T p.A475A Chr.1: 243505264-243505264 on Build GRCh38
278765 c.1355-1G>T p.? Chr.1: 243505335-243505335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756668340 Chr.1: 243505297-243505297 on Build GRCh38
rs892563259 Chr.1: 243505394-243505394 on Build GRCh38
rs771193056 Chr.1: 243505306-243505306 on Build GRCh38
rs552302039 Chr.1: 243505204-243505204 on Build GRCh38
rs746704800 Chr.1: 243505246-243505246 on Build GRCh38
rs762736239 Chr.1: 243505407-243505407 on Build GRCh38
rs765107759 Chr.1: 243505294-243505294 on Build GRCh38
rs201734360 Chr.1: 243505369-243505369 on Build GRCh38
rs587776935 Chr.1: 243505296-243505296 on Build GRCh38
rs753649886 Chr.1: 243505221-243505221 on Build GRCh38
rs918344063 Chr.1: 243505240-243505241 on Build GRCh38
rs565340176 Chr.1: 243505366-243505366 on Build GRCh38
rs778193336 Chr.1: 243505303-243505303 on Build GRCh38
rs751275167 Chr.1: 243505256-243505256 on Build GRCh38
rs766019860 Chr.1: 243505246-243505246 on Build GRCh38
rs765185824 Chr.1: 243505354-243505354 on Build GRCh38
rs749347952 Chr.1: 243505304-243505304 on Build GRCh38
rs1045371175 Chr.1: 243505349-243505349 on Build GRCh38
rs763992613 Chr.1: 243505209-243505209 on Build GRCh38
rs761768321 Chr.1: 243505225-243505225 on Build GRCh38
rs573383245 Chr.1: 243505288-243505288 on Build GRCh38
rs773093665 Chr.1: 243505361-243505361 on Build GRCh38
rs761558193 Chr.1: 243505337-243505337 on Build GRCh38
rs776919354 Chr.1: 243505355-243505355 on Build GRCh38
rs754658627 Chr.1: 243505384-243505384 on Build GRCh38
rs764600594 Chr.1: 243505239-243505239 on Build GRCh38
rs375329841 Chr.1: 243505331-243505331 on Build GRCh38
rs370750588 Chr.1: 243505240-243505240 on Build GRCh38
rs775103000 Chr.1: 243505209-243505213 on Build GRCh38
rs774657057 Chr.1: 243505311-243505311 on Build GRCh38
rs368205016 Chr.1: 243505334-243505334 on Build GRCh38
rs111322893 Chr.1: 243505377-243505379 on Build GRCh38
rs148332912 Chr.1: 243505273-243505273 on Build GRCh38
rs543291675 Chr.1: 243505242-243505242 on Build GRCh38
rs751148481 Chr.1: 243505378-243505378 on Build GRCh38
rs568959834 Chr.1: 243505374-243505376 on Build GRCh38
rs142844303 Chr.1: 243505330-243505330 on Build GRCh38
rs192105293 Chr.1: 243505205-243505205 on Build GRCh38
rs768201175 Chr.1: 243505193-243505193 on Build GRCh38
rs150573714 Chr.1: 243505291-243505291 on Build GRCh38
rs769005975 Chr.1: 243505245-243505246 on Build GRCh38

View Full Product Details