Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 176729578-176729803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VEGFC
Entrez Gene ID
7424
Gene Name
vascular endothelial growth factor C
Gene Aliases
Flt4-L, LMPH1D, VRP
UniGene
Hs.435215
Gene Chromosome Location
Chr.4: 176683534-176792745 on Build GRCh38
Transcripts
NM_005429.4

SNP Detail

SNP ID Chromosome Location
rs1006553027 Chr.4: 176729707-176729707 on Build GRCh38
rs775362388 Chr.4: 176729612-176729612 on Build GRCh38
rs184557768 Chr.4: 176729670-176729670 on Build GRCh38
rs1008209935 Chr.4: 176729677-176729677 on Build GRCh38
rs772222705 Chr.4: 176729608-176729608 on Build GRCh38
rs535467215 Chr.4: 176729718-176729718 on Build GRCh38
rs539876211 Chr.4: 176729695-176729695 on Build GRCh38
rs753815476 Chr.4: 176729747-176729747 on Build GRCh38
rs566826535 Chr.4: 176729713-176729713 on Build GRCh38
rs949316694 Chr.4: 176729619-176729619 on Build GRCh38
rs893547563 Chr.4: 176729701-176729701 on Build GRCh38
rs777500705 Chr.4: 176729739-176729739 on Build GRCh38
rs200219952 Chr.4: 176729722-176729722 on Build GRCh38
rs770236352 Chr.4: 176729663-176729663 on Build GRCh38
rs770289504 Chr.4: 176729768-176729768 on Build GRCh38
rs778894805 Chr.4: 176729753-176729753 on Build GRCh38
rs762308615 Chr.4: 176729633-176729633 on Build GRCh38
rs757082926 Chr.4: 176729752-176729752 on Build GRCh38
rs749178325 Chr.4: 176729757-176729757 on Build GRCh38
rs369596413 Chr.4: 176729624-176729624 on Build GRCh38
rs375440489 Chr.4: 176729756-176729756 on Build GRCh38
rs748341104 Chr.4: 176729763-176729763 on Build GRCh38
rs747267178 Chr.4: 176729620-176729620 on Build GRCh38
rs41278571 Chr.4: 176729712-176729712 on Build GRCh38
rs781587488 Chr.4: 176729762-176729762 on Build GRCh38
rs1030559639 Chr.4: 176729734-176729734 on Build GRCh38
rs41278569 Chr.4: 176729693-176729693 on Build GRCh38
rs750364363 Chr.4: 176729682-176729690 on Build GRCh38
rs1053514094 Chr.4: 176729683-176729683 on Build GRCh38
rs755922707 Chr.4: 176729735-176729735 on Build GRCh38
rs374947493 Chr.4: 176729755-176729755 on Build GRCh38
rs773392470 Chr.4: 176729770-176729770 on Build GRCh38
rs765928622 Chr.4: 176729698-176729698 on Build GRCh38
rs779772530 Chr.4: 176729748-176729749 on Build GRCh38
rs758131882 Chr.4: 176729713-176729714 on Build GRCh38
rs751092216 Chr.4: 176729702-176729702 on Build GRCh38
rs551187496 Chr.4: 176729627-176729627 on Build GRCh38
rs902373634 Chr.4: 176729630-176729630 on Build GRCh38
rs749126655 Chr.4: 176729605-176729605 on Build GRCh38

View Full Product Details