Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21671798-21672023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF9
Entrez Gene ID
2254
Gene Name
fibroblast growth factor 9
Gene Aliases
FGF-9, GAF, HBFG-9, HBGF-9, SYNS3
UniGene
Hs.111
Gene Chromosome Location
Chr.13: 21671076-21704501 on Build GRCh38
Transcripts
NM_002010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6333782 c.70G>T p.V24L Chr.13: 21671982-21671982 on Build GRCh38
4450991 c.100A>G p.S34G Chr.13: 21672012-21672012 on Build GRCh38
3417425 c.33C>A p.F11L Chr.13: 21671945-21671945 on Build GRCh38
1207082 c.20T>C p.V7A Chr.13: 21671932-21671932 on Build GRCh38
4046471 c.41A>G p.Q14R Chr.13: 21671953-21671953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049482815 Chr.13: 21671854-21671854 on Build GRCh38
rs767912297 Chr.13: 21671971-21671971 on Build GRCh38
rs770574167 Chr.13: 21671903-21671903 on Build GRCh38
rs752653250 Chr.13: 21671870-21671870 on Build GRCh38
rs900849065 Chr.13: 21671839-21671839 on Build GRCh38
rs761983668 Chr.13: 21671966-21671966 on Build GRCh38
rs780816251 Chr.13: 21671907-21671907 on Build GRCh38
rs565147519 Chr.13: 21671850-21671850 on Build GRCh38
rs576881958 Chr.13: 21672000-21672000 on Build GRCh38
rs759293888 Chr.13: 21671988-21671988 on Build GRCh38
rs182718810 Chr.13: 21671950-21671950 on Build GRCh38
rs886050044 Chr.13: 21671876-21671877 on Build GRCh38
rs1053235183 Chr.13: 21671827-21671827 on Build GRCh38
rs200508154 Chr.13: 21671902-21671902 on Build GRCh38
rs758229041 Chr.13: 21671873-21671873 on Build GRCh38
rs770172531 Chr.13: 21671945-21671945 on Build GRCh38
rs752566182 Chr.13: 21671875-21671876 on Build GRCh38
rs868605138 Chr.13: 21671928-21671928 on Build GRCh38
rs1037354014 Chr.13: 21671832-21671832 on Build GRCh38
rs145436564 Chr.13: 21671954-21671954 on Build GRCh38
rs750656784 Chr.13: 21671985-21671985 on Build GRCh38
rs892782413 Chr.13: 21671833-21671833 on Build GRCh38
rs746289906 Chr.13: 21671942-21671942 on Build GRCh38
rs774583506 Chr.13: 21671964-21671964 on Build GRCh38
rs763543332 Chr.13: 21671957-21671957 on Build GRCh38
rs1038941312 Chr.13: 21671852-21671852 on Build GRCh38
rs997306736 Chr.13: 21671955-21671955 on Build GRCh38
rs777367470 Chr.13: 21671881-21671881 on Build GRCh38
rs17069965 Chr.13: 21671900-21671900 on Build GRCh38
rs778769503 Chr.13: 21671868-21671868 on Build GRCh38
rs768992823 Chr.13: 21671960-21671960 on Build GRCh38

View Full Product Details