Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 164559730-164560003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PBX1
Entrez Gene ID
5087
Gene Name
PBX homeobox 1
Gene Aliases
-
UniGene
Hs.557097
Gene Chromosome Location
Chr.1: 164559360-164886047 on Build GRCh38
Transcripts
NM_001204961.1 NM_001204963.1 NM_002585.3 XM_005245228.3 XM_005245229.3 XM_011509590.1 XM_011509591.2 XM_011509592.1 XM_017001395.1 XM_017001396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7382532 c.167A>G p.Q56R Chr.1: 164559989-164559989 on Build GRCh38
5553357 c.107A>G p.E36G Chr.1: 164559929-164559929 on Build GRCh38
5780453 c.65T>G p.L22R Chr.1: 164559887-164559887 on Build GRCh38
5983413 c.61G>A p.G21S Chr.1: 164559883-164559883 on Build GRCh38
5553358 c.107A>G p.E36G Chr.1: 164559929-164559929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751494699 Chr.1: 164559893-164559893 on Build GRCh38
rs1000903065 Chr.1: 164559921-164559921 on Build GRCh38
rs770433143 Chr.1: 164559810-164559810 on Build GRCh38
rs375925928 Chr.1: 164559831-164559831 on Build GRCh38
rs747490519 Chr.1: 164559854-164559854 on Build GRCh38
rs763090532 Chr.1: 164559879-164559879 on Build GRCh38
rs372558581 Chr.1: 164559900-164559900 on Build GRCh38
rs970787385 Chr.1: 164559768-164559772 on Build GRCh38
rs111531180 Chr.1: 164559974-164559974 on Build GRCh38
rs2275558 Chr.1: 164559883-164559883 on Build GRCh38
rs144593424 Chr.1: 164559948-164559948 on Build GRCh38
rs1025052883 Chr.1: 164559767-164559768 on Build GRCh38
rs769041303 Chr.1: 164559872-164559872 on Build GRCh38
rs773577978 Chr.1: 164559878-164559878 on Build GRCh38
rs940224282 Chr.1: 164559852-164559852 on Build GRCh38
rs779217453 Chr.1: 164559815-164559816 on Build GRCh38
rs921304480 Chr.1: 164559799-164559799 on Build GRCh38
rs544057798 Chr.1: 164559874-164559874 on Build GRCh38
rs1057295149 Chr.1: 164559873-164559873 on Build GRCh38
rs958254448 Chr.1: 164559844-164559844 on Build GRCh38
rs558520686 Chr.1: 164559791-164559791 on Build GRCh38
rs370561885 Chr.1: 164559935-164559935 on Build GRCh38
rs1044664177 Chr.1: 164559909-164559909 on Build GRCh38
rs755790509 Chr.1: 164559772-164559772 on Build GRCh38
rs765608918 Chr.1: 164559877-164559877 on Build GRCh38
rs187406949 Chr.1: 164559814-164559814 on Build GRCh38
rs192264696 Chr.1: 164559914-164559914 on Build GRCh38
rs544814565 Chr.1: 164559773-164559773 on Build GRCh38
rs777341799 Chr.1: 164559956-164559956 on Build GRCh38
rs1012363761 Chr.1: 164559763-164559763 on Build GRCh38
rs1031011831 Chr.1: 164559834-164559834 on Build GRCh38
rs931299581 Chr.1: 164559811-164559811 on Build GRCh38

View Full Product Details