Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21671960-21672229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF9
Entrez Gene ID
2254
Gene Name
fibroblast growth factor 9
Gene Aliases
FGF-9, GAF, HBFG-9, HBGF-9, SYNS3
UniGene
Hs.111
Gene Chromosome Location
Chr.13: 21671076-21704501 on Build GRCh38
Transcripts
NM_002010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3671250 c.121G>A p.E41K Chr.13: 21672033-21672033 on Build GRCh38
6138968 c.244A>G p.I82V Chr.13: 21672156-21672156 on Build GRCh38
7118381 c.251G>A p.G84E Chr.13: 21672163-21672163 on Build GRCh38
1743345 c.177G>A p.G59G Chr.13: 21672089-21672089 on Build GRCh38
5531254 c.166C>T p.H56Y Chr.13: 21672078-21672078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774988398 Chr.13: 21672063-21672063 on Build GRCh38
rs145719719 Chr.13: 21672002-21672002 on Build GRCh38
rs576881958 Chr.13: 21672000-21672000 on Build GRCh38
rs755802170 Chr.13: 21672150-21672150 on Build GRCh38
rs779607710 Chr.13: 21672183-21672183 on Build GRCh38
rs780871403 Chr.13: 21672037-21672037 on Build GRCh38
rs756799366 Chr.13: 21672144-21672144 on Build GRCh38
rs759293888 Chr.13: 21671988-21671988 on Build GRCh38
rs1004951478 Chr.13: 21672068-21672068 on Build GRCh38
rs749722061 Chr.13: 21672175-21672175 on Build GRCh38
rs1016708024 Chr.13: 21672131-21672131 on Build GRCh38
rs541049028 Chr.13: 21672071-21672071 on Build GRCh38
rs751136069 Chr.13: 21672024-21672024 on Build GRCh38
rs267603776 Chr.13: 21672088-21672088 on Build GRCh38
rs766456148 Chr.13: 21672077-21672077 on Build GRCh38
rs755561237 Chr.13: 21672066-21672066 on Build GRCh38
rs748699086 Chr.13: 21672198-21672198 on Build GRCh38
rs751475837 Chr.13: 21672138-21672138 on Build GRCh38
rs756744215 Chr.13: 21672030-21672030 on Build GRCh38
rs745487908 Chr.13: 21672047-21672047 on Build GRCh38
rs767042516 Chr.13: 21672145-21672145 on Build GRCh38
rs750656784 Chr.13: 21671985-21671985 on Build GRCh38
rs780484804 Chr.13: 21672054-21672054 on Build GRCh38
rs769207038 Chr.13: 21672062-21672062 on Build GRCh38
rs779502534 Chr.13: 21672174-21672174 on Build GRCh38
rs376187337 Chr.13: 21672003-21672003 on Build GRCh38
rs150678654 Chr.13: 21672025-21672025 on Build GRCh38
rs769287069 Chr.13: 21672053-21672053 on Build GRCh38
rs749883098 Chr.13: 21672148-21672148 on Build GRCh38
rs755277615 Chr.13: 21672178-21672178 on Build GRCh38
rs775361742 Chr.13: 21672104-21672104 on Build GRCh38
rs764075237 Chr.13: 21672122-21672122 on Build GRCh38
rs749555659 Chr.13: 21672059-21672059 on Build GRCh38
rs772251822 Chr.13: 21672067-21672067 on Build GRCh38
rs760974340 Chr.13: 21672074-21672074 on Build GRCh38
rs762649405 Chr.13: 21672119-21672119 on Build GRCh38
rs570227715 Chr.13: 21672008-21672008 on Build GRCh38
rs758207436 Chr.13: 21672006-21672008 on Build GRCh38

View Full Product Details