Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 176687310-176687581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VEGFC
Entrez Gene ID
7424
Gene Name
vascular endothelial growth factor C
Gene Aliases
Flt4-L, LMPH1D, VRP
UniGene
Hs.435215
Gene Chromosome Location
Chr.4: 176683534-176792745 on Build GRCh38
Transcripts
NM_005429.4

SNP Detail

SNP ID Chromosome Location
rs7664413 Chr.4: 176687553-176687553 on Build GRCh38
rs748396337 Chr.4: 176687363-176687363 on Build GRCh38
rs748304588 Chr.4: 176687483-176687483 on Build GRCh38
rs923653602 Chr.4: 176687427-176687427 on Build GRCh38
rs768075524 Chr.4: 176687470-176687470 on Build GRCh38
rs768004289 Chr.4: 176687531-176687534 on Build GRCh38
rs369537352 Chr.4: 176687433-176687433 on Build GRCh38
rs768802006 Chr.4: 176687358-176687358 on Build GRCh38
rs757091711 Chr.4: 176687441-176687441 on Build GRCh38
rs763799312 Chr.4: 176687434-176687434 on Build GRCh38
rs774825708 Chr.4: 176687440-176687440 on Build GRCh38
rs902478510 Chr.4: 176687344-176687344 on Build GRCh38
rs371693961 Chr.4: 176687442-176687442 on Build GRCh38
rs778948257 Chr.4: 176687345-176687345 on Build GRCh38
rs930222184 Chr.4: 176687556-176687556 on Build GRCh38
rs372436876 Chr.4: 176687367-176687367 on Build GRCh38
rs749474797 Chr.4: 176687524-176687524 on Build GRCh38
rs745769667 Chr.4: 176687351-176687351 on Build GRCh38
rs957771026 Chr.4: 176687381-176687381 on Build GRCh38
rs267600102 Chr.4: 176687503-176687503 on Build GRCh38
rs753220179 Chr.4: 176687502-176687502 on Build GRCh38
rs778194080 Chr.4: 176687507-176687507 on Build GRCh38
rs771362030 Chr.4: 176687528-176687528 on Build GRCh38
rs772509359 Chr.4: 176687545-176687545 on Build GRCh38
rs766634395 Chr.4: 176687400-176687400 on Build GRCh38
rs774948307 Chr.4: 176687401-176687401 on Build GRCh38
rs575984035 Chr.4: 176687340-176687340 on Build GRCh38
rs866628334 Chr.4: 176687445-176687445 on Build GRCh38
rs759923423 Chr.4: 176687412-176687412 on Build GRCh38
rs759858667 Chr.4: 176687462-176687462 on Build GRCh38
rs202208990 Chr.4: 176687380-176687380 on Build GRCh38
rs755693482 Chr.4: 176687337-176687337 on Build GRCh38
rs999139449 Chr.4: 176687371-176687371 on Build GRCh38
rs780029765 Chr.4: 176687476-176687476 on Build GRCh38
rs879072706 Chr.4: 176687352-176687352 on Build GRCh38
rs775968386 Chr.4: 176687549-176687549 on Build GRCh38
rs375957104 Chr.4: 176687540-176687540 on Build GRCh38
rs753457751 Chr.4: 176687438-176687438 on Build GRCh38
rs985631564 Chr.4: 176687387-176687387 on Build GRCh38
rs767056045 Chr.4: 176687426-176687426 on Build GRCh38
rs1039688099 Chr.4: 176687535-176687535 on Build GRCh38
rs564667848 Chr.4: 176687472-176687472 on Build GRCh38
rs769963677 Chr.4: 176687491-176687491 on Build GRCh38
rs763455051 Chr.4: 176687399-176687399 on Build GRCh38
rs745847895 Chr.4: 176687452-176687452 on Build GRCh38
rs774509633 Chr.4: 176687538-176687538 on Build GRCh38
rs776509676 Chr.4: 176687361-176687361 on Build GRCh38

View Full Product Details