Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145979.2 | NP_666091.1 | 15 | 2450 | |
GenBank mRNA | AK034549.1 | - | 10 | 1531 | |
AK147695.1 | - | 15 | 2594 | ||
AK153826.1 | - | 8 | 1208 | ||
AK220473.1 | - | 14 | 2358 | ||
BC058578.1 | - | 15 | 2450 | 6489649 | |
BC059082.1 | - | 2 | 231 |