Gene Transcripts

Gene Symbol
Psen2
Entrez Gene ID
19165
Gene Name
presenilin 2
Gene Aliases
ALG-3, Ad4h, Alg3, PS-2, PS2, Psnl2, STM2
Chromosome Location
Chr. 1: 180227004 - 180263476 on Build GRCm38
UniGene ID
Mm.330850
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128605.1 NP_001122077.1 3 402
NM_011183.3 NP_035313.2 3 411
GenBank mRNA AF038935.1 - 2 339
AK014706.1 - 2 346
AK148111.1 - 3 403
AK154119.1 - 3 347
BC010403.1 - 2 357 4038761
U57324.1 - 3 362

View Full Product Details