Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177719.1 | NP_001171190.1 | 17 | 3076 | |
NM_001177720.1 | NP_001171191.1 | 15 | 2764 | ||
NM_145527.4 | NP_663502.3 | 16 | 3016 | ||
GenBank mRNA | AK035777.1 | - | 17 | 3057 | |
AK036347.1 | - | 16 | 3017 | ||
AK122257.1 | - | 17 | 2918 | ||
AY263980.1 | - | 17 | 2894 | ||
AY263981.1 | - | 17 | 2894 | ||
AY263982.1 | - | 16 | 2834 | ||
AY263983.1 | - | 17 | 2837 | ||
AY263984.1 | - | 16 | 2777 | ||
AY263985.1 | - | 16 | 2759 | ||
AY263986.1 | - | 16 | 2759 | ||
AY263987.1 | - | 16 | 2759 | ||
AY263988.1 | - | 16 | 2759 | ||
AY263989.1 | - | 16 | 2759 | ||
BC042212.1 | - | 6 | 754 | ||
BC063386.1 | - | 16 | 2817 | 5401819 |