Gene Transcripts

Gene Symbol
Setd2
Entrez Gene ID
235626
Gene Name
SET domain containing 2
Gene Aliases
4921524K10Rik, BC031601, KMT3A
Chromosome Location
Chr. 9: 110529296 - 110618633 on Build GRCm38
UniGene ID
Mm.288949
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081340.2 NP_001074809.2 12 5603
GenBank mRNA AK046730.1 - 1 108
AK173246.1 - 1 24
BC059049.1 - 1 126
CX564178.1 - 4 509

View Full Product Details