Gene Transcripts
- Gene Symbol
- Sec61a1
- Entrez Gene ID
- 53421
- Gene Name
- Sec61 alpha 1 subunit (S. cerevisiae)
- Gene Aliases
- AA408394, AA410007, Sec61a
- Chromosome Location
- Chr. 6: 88503607 - 88518800 on Build GRCm38
- UniGene ID
- Mm.28375
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016906.4 |
NP_058602.1
|
2, 3 |
180 |
|
GenBank mRNA |
AB032902.1 |
- |
2, 3 |
192 |
|
AF145253.1 |
- |
2, 3 |
102 |
|
AF222743.1 |
- |
2, 3 |
102 |
|
AK088473.1 |
- |
2, 3 |
181 |
|
AK153055.1 |
- |
2, 3 |
170 |
|
AK159349.1 |
- |
2, 3 |
168 |
|
AK167484.1 |
- |
2, 3 |
110 |
|
AK219834.1 |
- |
1, 2 |
23 |
|
BC003707.1 |
- |
2, 3 |
106 |
3592772 |
View Full Product Details