Gene Transcripts
- Gene Symbol
- Gm5608
- Entrez Gene ID
- 434282
- Gene Name
- predicted gene 5608
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK089977.1 |
- |
1, 2 |
711 |
|
View Full Product Details